Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729522_s1
          See other OLFR891 GE Assays ›
          Gene Symbol:
          Olfr891
          Gene Name:
          olfactory receptor 891
          Gene Aliases:
          MOR170-9
          Chromosome Location:
          Chr.9: 38179871 - 38180821 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729522_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr891
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146478.2 NP_666689.2 1 855 153

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr891

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977382 NM_146478.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159976 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134887 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154689 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977382 NM_146478.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P575980 NM_146478.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433880 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134887 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154689 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159976 - -
          Whole Mouse Genome G4122A A_52_P575980 NM_146478.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139002 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154689 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159976 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134887 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07773624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07773620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07755517 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07755525 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07755526 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07773616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07853391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07494748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07886721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433868 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592370 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592369 - -
          Affymetrix Mouse Clariom S Array PSR0900017171.mm.1 NM_146478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017171.mm.1 NM_146478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017171.mm.1 TC0900002059.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 460056 BC029168.1 genbank_mrna_mouse
          MouseWG-6 v2 Expression BeadChip 460056 BC039918.1 genbank_mrna_mouse
          Illumina mouse-6 v1.0 expression beadchip 460056 BC029168.1 genbank_mrna_mouse
          Illumina mouse-6 v1.0 expression beadchip 460056 BC039918.1 genbank_mrna_mouse
          Illumina mouseRef-8 v1.0 expression beadchip 0000460056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2689169 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2689169 - -
          mouse-6 v1.1 (Illumina) scl36019.3.332_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 460056 BC029168.1 genbank_mrna_mouse
          Illumina Sentrix mouse-6.v1 bead array 460056 BC039918.1 genbank_mrna_mouse
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36019.3.332_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460056 BC029168.1 genbank_mrna_mouse
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460056 BC039918.1 genbank_mrna_mouse
          MouseRef-8 v2 Expression BeadChip ILMN_1233711 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233711 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline