Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729979_s1
          See other OLFR1361 GE Assays ›
          Gene Symbol:
          Olfr1361
          Gene Name:
          olfactory receptor 1361
          Gene Aliases:
          MOR256-12
          Chromosome Location:
          Chr.13: 21658368 - 21664523 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729979_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1361
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146541.2 NP_666752.1 1 642 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1361

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976834 NM_146541.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148915 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976834 NM_146541.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P103850 NM_146541.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28873935 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148915 - -
          Whole Mouse Genome G4122A A_52_P103850 NM_146541.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116840 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10821484 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10582295 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10582302 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11042971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28873910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10973476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10582305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10761483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10973484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10761489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10761488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28873922 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10408067 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408066 - -
          Affymetrix Mouse Clariom S Array PSR1300009824.mm.1 NM_146541.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009824.mm.1 NM_146541.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009824.mm.1 TC1300001553.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 789244 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5860148 - -
          Illumina mouse-6 v1.0 expression beadchip 5860148 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005860148 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2643367 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2643367 - -
          mouse-6 v1.1 (Illumina) scl11532.1.1_186-S - -
          Illumina Sentrix mouse-6.v1 bead array 5860148 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl11532.1.1_186-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5860148 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2643367 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2643367 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline