Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00772826_m1
          See other KBTBD3 GE Assays ›
          Gene Symbol:
          Kbtbd3
          Gene Name:
          kelch repeat and BTB (POZ) domain containing 3
          Gene Aliases:
          2200003A07Rik, Bklhd3
          Chromosome Location:
          Chr.9: 4309743 - 4331829 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00772826_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kbtbd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164574.1 NP_001158046.1 2 427 94
          NM_026962.3 NP_081238.2 3 459 94
          GenBank mRNA AK008627.1 - 2 317 94
          AK034334.1 - 3 367 94
          AK078744.1 - 2 339 94
          AK083497.1 - 3 441 94
          AK168345.1 - 2 273 94
          BB264869.2 - 2 428 94
          BC024938.1 - 2 323 94
          BC027392.1 - 3 363 4954712 94
          BE650264.1 - 3 460 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000025.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kbtbd3

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26276397 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965709 BC027392.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965709 AK034334.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965709 AK078744.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965709 AK083497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965709 NM_001164574.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 617275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79999 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 72309 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115735 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104510 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965709 BC027392.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965709 AK034334.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965709 AK078744.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965709 AK083497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965709 NM_001164574.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26276400 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115735 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06688059 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104510 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06688042 - -
          Whole Mouse Genome G4122A A_52_P574081 BC027392.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574081 AK168345.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574081 BE650264.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574081 BC024938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574081 AK034334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593986 BC027392.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593986 AK034334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593986 AK078744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593986 AK083497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593986 NM_001164574.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130332 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104510 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115735 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07680787 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07680792 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07680785 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26276397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26276457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07456256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07680785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08151424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07570503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07680791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07845749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07655069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26276505 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10582976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582974 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582975 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582977 - -
          Affymetrix HT MG-430 PM Array Plate 1439062_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10582973 - -
          Mouse Genome 430 2.0 Array 1439062_PM_at - -
          Mouse Genome 430 2.0 Array 1439062_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69149_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69149_at - -
          Affymetrix Mouse Clariom S Array PSR0900000194.mm.1 NM_001164574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000194.mm.1 NM_001164574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000194.mm.1 NM_026962.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000194.mm.1 NM_026962.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000194.mm.1 TC0900000025.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000193.mm.1 NM_001164574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000193.mm.1 NM_001164574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000193.mm.1 TC0900000025.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000197.mm.1 NM_026962.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000197.mm.1 NM_026962.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000197.mm.1 TC0900000025.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000199.mm.1 NM_001164574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000199.mm.1 NM_001164574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000199.mm.1 NM_026962.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000199.mm.1 NM_026962.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000199.mm.1 TC0900000025.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000198.mm.1 NM_001164574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000198.mm.1 NM_001164574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000198.mm.1 NM_026962.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000198.mm.1 NM_026962.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000198.mm.1 TC0900000025.mm.1 MTA TC
          Mouse Expression Set 430 1439062_at - -
          Mouse Expression Set 430 1439062_PM_at - -
          Murine Genome U74v2 Set 168575_r_at - -
          Murine Genome U74v2 Set 168575_PM_r_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 759036 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3360711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360711 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214947 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214947 - -
          mouse-6 v1.1 (Illumina) scl37320.5_187-S - -
          Illumina Sentrix mouse-6.v1 bead array 3360711 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37320.5_187-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1214947 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214947 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline