Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00777952_s1
          See other ZFP119A GE Assays ›
          Gene Symbol:
          Zfp119a
          Gene Name:
          zinc finger protein 119a
          Gene Aliases:
          AL024077, Mzf13, Zfp119
          Chromosome Location:
          Chr.17: 55864886 - 55879685 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00777952_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp119a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144546.6 NP_653129.1 4 1764 158
          GenBank mRNA AF242376.1 - 4 1765 158
          AK047498.1 - 804 158
          BC118950.2 - 4 1741 40092426 158

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp119a

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P372092 AK047498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P372092 BC118950.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P372092 NM_144546.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P372092 AF242376.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P372092 AK047498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P372092 BC118950.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P372092 NM_144546.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P372092 AF242376.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842147 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 250335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659678 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771541 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937273 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 110414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159906 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11767 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148085 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171418 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P372092 AK047498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P372092 BC118950.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P372092 NM_144546.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P372092 AF242376.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P372092 AK047498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P372092 BC118950.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P372092 NM_144546.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P372092 AF242376.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P220049 AK047498.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31252753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31252732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31252730 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03338629 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08809891 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08809860 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148085 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08809854 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08809906 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08809901 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171418 - -
          Whole Mouse Genome G4122A A_52_P1124147 - -
          Whole Mouse Genome G4122A A_51_P220049 AK047498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372092 AK047498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372092 BC118950.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372092 NM_144546.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372092 AF242376.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372092 AK047498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372092 BC118950.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372092 NM_144546.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372092 AF242376.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111436 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08809906 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08809860 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171418 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148085 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08809899 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13409833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31252710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13558990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13057868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13117474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31252832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10451880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451881 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451883 - -
          Affymetrix HT MG-430 PM Array Plate 1425010_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10451879 - -
          Mouse Genome 430 2.0 Array 1425010_at - -
          Mouse Genome 430 2.0 Array 1425010_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 104349_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 104349_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700021496.mm.1 NM_144546.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021496.mm.1 TC1700002303.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1425010_at - -
          Affymetrix HT Mouse Genome 430A Array 1425010_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 441051 - -
          ABI Mouse Genome Survey Microarray 445933 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1090731 - -
          Illumina mouse-6 v1.0 expression beadchip 1240551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001240551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090731 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2520569 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2431067 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2520569 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2431067 - -
          mouse-6 v1.1 (Illumina) scl0104349.1_226-S - -
          mouse-6 v1.1 (Illumina) scl00104349.2_320-S - -
          Illumina Sentrix mouse-6.v1 bead array 1240551 - -
          Illumina Sentrix mouse-6.v1 bead array 1090731 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00104349.2_320-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0104349.1_226-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090731 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2907989 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2431067 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2907989 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2431067 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline