Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00780551_s1
          See other RRP7A GE Assays ›
          Gene Symbol:
          Rrp7a
          Gene Name:
          ribosomal RNA processing 7 homolog A (S. cerevisiae)
          Gene Aliases:
          1110014J01Rik, AA408146, Kheg1
          Chromosome Location:
          Chr.15: 83115846 - 83122826 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00780551_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rrp7a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK032596.1 - 4052 145
          AK036247.1 - 4034 145

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Mus musculus 1700 Inventoried

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30386715 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30386711 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30386713 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30386709 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150014 AK036247.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150014 AK032596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150014 AK036247.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150014 AK032596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2108206 CO042481.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952537 AK168850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952537 BC012523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952537 AK153565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952537 AK159553.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952537 AK036247.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694013 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572760 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 119960 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116769 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133637 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2108206 CO042481.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952537 AK168850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952537 BC012523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952537 AK153565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952537 AK159553.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952537 AK036247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150014 AK036247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150014 AK032596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150014 AK036247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150014 AK032596.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30386711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30386704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30386713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30386719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30386721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30386717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30386715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30386702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30386712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30386708 - -
          Whole Mouse Genome G4122A A_51_P150014 AK036247.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150014 AK032596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150014 AK036247.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150014 AK032596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361906 AK168850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361906 BC012523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361906 AK153565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361906 AK159553.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361906 AK036247.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137107 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12306665 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11919516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30386709 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30386715 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30386711 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30386713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30386694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11919524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12329385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11920877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12320310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11850787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12306665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12320294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12329375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12329381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30386717 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430942 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425794 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425792 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430943 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430944 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425793 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425791 - -
          Affymetrix HT MG-430 PM Array Plate 1444383_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1448141_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456814_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10430941 - -
          Affymetrix Mouse Gene 1.1 ST Array 10425781 - -
          Mouse Genome 430 2.0 Array 1456814_PM_at - -
          Mouse Genome 430 2.0 Array 1448141_PM_at - -
          Mouse Genome 430 2.0 Array 1444383_PM_at - -
          Mouse Genome 430 2.0 Array 1456814_at - -
          Mouse Genome 430 2.0 Array 1444383_at - -
          Mouse Genome 430 2.0 Array 1448141_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74778_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74778_PM_at - -
          Mouse Expression Set 430 1448141_at - -
          Mouse Expression Set 430 1448141_PM_at - -
          Mouse Expression Set 430 1456814_PM_at - -
          Mouse Expression Set 430 1444383_PM_at - -
          Mouse Expression Set 430 1456814_at - -
          Mouse Expression Set 430 1444383_at - -
          Murine Genome U74v2 Set 160382_at - -
          Murine Genome U74v2 Set 160382_PM_at - -
          Mouse Genome 430A 2.0 Array 1448141_at - -
          Mouse Genome 430A 2.0 Array 1448141_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448141_at - -
          Affymetrix HT Mouse Genome 430A Array 1448141_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444383_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456814_at - -
          Affymetrix HT Mouse Genome 430B Array 1444383_at - -
          Affymetrix HT Mouse Genome 430B Array 1456814_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 859193 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1980332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213124 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213124 - -
          mouse-6 v1.1 (Illumina) scl46903.8_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46903.8_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1213124 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213124 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0