Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00782974_s1
          See other MRPL57 GE Assays ›
          Gene Symbol:
          Mrpl57
          Gene Name:
          mitochondrial ribosomal protein L53
          Gene Aliases:
          2610025P17Rik, AV005697, Mrp63
          Chromosome Location:
          Chr.14: 57826228 - 57829087 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00782974_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl57
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026401.2 NP_080677.1 2 2253 136
          GenBank mRNA AK011558.1 - 2 2254 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl57

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29630114 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29630115 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29630113 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P528707 BC049555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P528707 BC055723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P528707 NM_026401.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P528707 AB049958.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P528707 AK003130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P528707 BC049555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P528707 BC055723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P528707 NM_026401.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P528707 AB049958.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P528707 AK003130.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577414 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140778 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P528707 BC049555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P528707 BC055723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P528707 NM_026401.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P528707 AB049958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P528707 AK003130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P528707 BC049555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P528707 BC055723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P528707 NM_026401.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P528707 AB049958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P528707 AK003130.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P168219 NM_026401.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P168219 AK011558.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29630137 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140778 - -
          Whole Mouse Genome G4122A A_51_P168219 NM_026401.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P168219 AK011558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P528707 BC049555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P528707 BC055723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P528707 NM_026401.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P528707 AB049958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P528707 AK003130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P528707 BC049555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P528707 BC055723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P528707 NM_026401.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P528707 AB049958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P528707 AK003130.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128612 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140778 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11563094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11431014 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11431012 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11431017 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29630115 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29630114 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29630113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29630113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11431012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11453422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11654065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29630135 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10415648 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415646 - -
          Affymetrix HT MG-430 PM Array Plate 1460225_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418137_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10415645 - -
          Mouse Genome 430 2.0 Array 1460225_at - -
          Mouse Genome 430 2.0 Array 1418137_at - -
          Mouse Genome 430 2.0 Array 1418137_PM_at - -
          Mouse Genome 430 2.0 Array 1460225_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67840_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67840_PM_at - -
          Mouse Expression Set 430 1460225_PM_at - -
          Mouse Expression Set 430 1418137_PM_at - -
          Mouse Expression Set 430 1460225_at - -
          Mouse Expression Set 430 1418137_at - -
          Murine Genome U74v2 Set 96931_PM_at - -
          Murine Genome U74v2 Set 96931_at - -
          Murine Genome U74v2 Set 134180_at - -
          Murine Genome U74v2 Set 134180_PM_at - -
          Mouse Genome 430A 2.0 Array 1418137_PM_at - -
          Mouse Genome 430A 2.0 Array 1460225_PM_at - -
          Mouse Genome 430A 2.0 Array 1418137_at - -
          Mouse Genome 430A 2.0 Array 1460225_at - -
          Affymetrix HT Mouse Genome 430A Array 1460225_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418137_at - -
          Affymetrix HT Mouse Genome 430A Array 1418137_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460225_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 517773 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2360193 - -
          Illumina mouse-6 v1.0 expression beadchip 2360193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002360193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258748 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258748 - -
          mouse-6 v1.1 (Illumina) scl46237.2_415-S - -
          Illumina Sentrix mouse-6.v1 bead array 2360193 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46237.2_415-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360193 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2790113 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2790113 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline