Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00803242_m1
          See other RAB19 GE Assays ›
          Gene Symbol:
          Rab19
          Gene Name:
          RAB19, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 39381428 - 39390666 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00803242_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011226.1 NP_035356.1 3 - 4 479 58
          GenBank mRNA AB232613.1 - 2 - 3 385 58
          AK017590.1 - 3 - 4 513 58
          BC032936.1 - 3 - 4 479 1379043 58
          X80473.1 - 2 - 3 385 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000392.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab19

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          small GTPase mediated signal transduction
          protein transport
          Rab protein signal transduction
          nucleotide binding
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414126 BC032936.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414126 AK017590.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414126 NM_011226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414126 BC032936.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414126 AK017590.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414126 NM_011226.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439138 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124490 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139090 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414126 BC032936.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414126 AK017590.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414126 NM_011226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414126 BC032936.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414126 AK017590.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414126 NM_011226.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414126 BC032936.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414126 AK017590.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414126 NM_011226.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414126 BC032936.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414126 AK017590.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414126 NM_011226.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24382823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24382816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24382826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24382828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24382819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24382829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24382821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24382813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24382817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24382825 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139090 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05871251 - -
          Whole Mouse Genome G4122A A_51_P414126 BC032936.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414126 AK017590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414126 NM_011226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414126 BC032936.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414126 AK017590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414126 NM_011226.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102675 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139090 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05691261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05691260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24382813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05691270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05098975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05691260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05757255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05807566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05438281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05115505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05454640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05365207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05691271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05757254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05744130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05060099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05627260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05691263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24382823 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537435 - -
          Affymetrix HT MG-430 PM Array Plate 1430148_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10537428 - -
          Mouse Genome 430 2.0 Array 1430148_PM_at - -
          Mouse Genome 430 2.0 Array 1430148_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19331_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19331_at - -
          Affymetrix Mouse Clariom S Array PSR0600003031.mm.1 TC0600000392.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003031.mm.1 NM_011226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003031.mm.1 NM_011226.1 Refseq
          Mouse Expression Set 430 1430148_at - -
          Mouse Expression Set 430 1430148_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430148_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430148_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 746252 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5570035 - -
          Illumina mouse-6 v1.0 expression beadchip 5570035 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570035 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605960 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605960 - -
          mouse-6 v1.1 (Illumina) scl30175.4.1_38-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570035 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30175.4.1_38-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570035 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605960 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2942082 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2942082 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605960 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline