Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00804065_m1
          See other RNF187 GE Assays ›
          Gene Symbol:
          Rnf187
          Gene Name:
          ring finger protein 187
          Gene Aliases:
          2410016F01Rik, AB030190, RACO-1
          Chromosome Location:
          Chr.11: 58932288 - 58938906 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00804065_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf187
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022423.2 NP_071868.2 1 - 2 509 85
          GenBank mRNA AB030190.1 - 1 - 2 501 85
          AK037991.1 - 1 - 2 424 85
          AK075944.1 - 1 - 2 520 85
          AK076158.1 - 1 - 2 499 85
          AK088973.1 - 1 - 2 512 85
          BC072613.1 - 1 - 2 502 85
          BC096522.1 - 1 - 2 473 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002836.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf187

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of transcription, DNA-templated
          protein autoubiquitination
          protein K48-linked ubiquitination
          ubiquitin-protein transferase activity
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518298 AK219828.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518298 BC072613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518298 AK076158.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518298 AB030190.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518298 AK037991.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518298 AK219828.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518298 BC072613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518298 AK076158.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518298 AB030190.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518298 AK037991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518298 AK219828.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518298 BC072613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518298 AK076158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518298 AB030190.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518298 AK037991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518298 AK219828.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518298 BC072613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518298 AK076158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518298 AB030190.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518298 AK037991.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07358530 - -
          Whole Mouse Genome G4122A A_51_P518298 AK219828.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518298 BC072613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518298 AK076158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518298 AB030190.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518298 AK037991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518298 AK219828.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518298 BC072613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518298 AK076158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518298 AB030190.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518298 AK037991.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833620 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10386234 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386231 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386233 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386232 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386235 - -
          Affymetrix HT MG-430 PM Array Plate 1423897_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423896_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1451157_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10386230 - -
          Mouse Genome 430 2.0 Array 1451157_at - -
          Mouse Genome 430 2.0 Array 1423897_at - -
          Mouse Genome 430 2.0 Array 1423897_PM_at - -
          Mouse Genome 430 2.0 Array 1423896_PM_a_at - -
          Mouse Genome 430 2.0 Array 1451157_PM_at - -
          Mouse Genome 430 2.0 Array 1423896_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108660_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108660_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100025607.mm.1 NM_022423.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025607.mm.1 NM_022423.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025607.mm.1 TC1100002836.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025606.mm.1 NM_022423.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025606.mm.1 NM_022423.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025606.mm.1 TC1100002836.mm.1 MTA TC
          Mouse Expression Set 430 1423896_a_at - -
          Mouse Expression Set 430 1423897_PM_at - -
          Mouse Expression Set 430 1451157_at - -
          Mouse Expression Set 430 1423896_PM_a_at - -
          Mouse Expression Set 430 1451157_PM_at - -
          Mouse Expression Set 430 1423897_at - -
          Mouse Genome 430A 2.0 Array 1423896_a_at - -
          Mouse Genome 430A 2.0 Array 1451157_at - -
          Mouse Genome 430A 2.0 Array 1451157_PM_at - -
          Mouse Genome 430A 2.0 Array 1423897_at - -
          Mouse Genome 430A 2.0 Array 1423896_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1423897_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451157_at - -
          Affymetrix HT Mouse Genome 430A Array 1423896_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423897_at - -
          Affymetrix HT Mouse Genome 430A Array 1423897_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451157_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423896_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 904921 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1940519 - -
          Illumina mouse-6 v1.0 expression beadchip 1940519 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001940519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229770 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229770 - -
          mouse-6 v1.1 (Illumina) scl40177.4_172-S - -
          Illumina Sentrix mouse-6.v1 bead array 1940519 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40177.4_172-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1940519 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2701339 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40177.4_172-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline