| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900019691.mm.1 |
NM_183111.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019691.mm.1 |
NM_183111.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019691.mm.1 |
TC0900002345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900019694.mm.1 |
NM_183111.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019694.mm.1 |
NM_183111.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019694.mm.1 |
TC0900002345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900019690.mm.1 |
NM_183111.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019690.mm.1 |
NM_183111.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019690.mm.1 |
TC0900002345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900019693.mm.1 |
NM_183111.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019693.mm.1 |
NM_183111.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019693.mm.1 |
TC0900002345.mm.1 |
MTA TC |