Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805872_m1
          See other NRDE2 GE Assays ›
          Gene Symbol:
          Nrde2
          Gene Name:
          nrde-2 necessary for RNA interference, domain containing
          Gene Aliases:
          6720454P05Rik
          Chromosome Location:
          Chr.12: 100125450 - 100159931 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805872_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrde2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290303.1 NP_001277232.1 10 - 11 2306 78
          NM_183155.3 NP_898978.2 10 - 11 2291 78
          GenBank mRNA BC025577.1 - 5 - 6 911 5323598 78
          BC051175.1 - 9 - 10 2196 78
          DN174402.1 - 1 - 2 301 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002220.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nrde2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P213592 BC051175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P213592 BC002230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P213592 BC025577.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P213592 NM_183155.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P213592 BC051175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P213592 BC002230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P213592 BC025577.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P213592 NM_183155.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753772 BC134373.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753772 NM_146548.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507067 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320999 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736888 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883296 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 705396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223411 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 115560 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159319 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P213592 BC051175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P213592 BC002230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P213592 BC025577.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P213592 NM_183155.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P213592 BC051175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P213592 BC002230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P213592 BC025577.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P213592 NM_183155.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07724617 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02588795 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07724595 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159319 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02588787 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07724634 - -
          Whole Mouse Genome G4122A A_51_P213592 BC051175.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P213592 BC002230.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P213592 BC025577.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P213592 NM_183155.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P213592 BC051175.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P213592 BC002230.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P213592 BC025577.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P213592 NM_183155.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02588795 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09926213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10157744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10362116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09926218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09793173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10374353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10392931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10216954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09820765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09949781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09926216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10041063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10326538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09921907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10429843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09917844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09926205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10326537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10092178 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10402085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402086 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402093 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402087 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402095 - -
          Affymetrix HT MG-430 PM Array Plate 1441426_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451303_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10402080 - -
          Mouse Genome 430 2.0 Array 1441426_PM_at - -
          Mouse Genome 430 2.0 Array 1451303_at - -
          Mouse Genome 430 2.0 Array 1451303_PM_at - -
          Mouse Genome 430 2.0 Array 1441426_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217827_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217827_at - -
          Affymetrix Mouse Clariom S Array PSR1200015289.mm.1 NM_183155.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015289.mm.1 TC1200002220.mm.1 MTA TC
          Mouse Expression Set 430 1451303_PM_at - -
          Mouse Expression Set 430 1451303_at - -
          Murine Genome U74v2 Set 130739_PM_at - -
          Murine Genome U74v2 Set 130739_at - -
          Mouse Genome 430A 2.0 Array 1451303_PM_at - -
          Mouse Genome 430A 2.0 Array 1451303_at - -
          Affymetrix HT Mouse Genome 430A Array 1451303_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451303_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 695846 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106380102 - -
          MouseWG-6 v2 Expression BeadChip 430746 - -
          Illumina mouse-6 v1.0 expression beadchip 430746 - -
          Illumina mouse-6 v1.0 expression beadchip 106380102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430746 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248444 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238819 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248444 - -
          mouse-6 v1.1 (Illumina) scl0217827.2_0-S - -
          mouse-6 v1.1 (Illumina) scl0002335.1_101-S - -
          Illumina Sentrix mouse-6.v1 bead array 106380102 - -
          Illumina Sentrix mouse-6.v1 bead array 430746 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0217827.2_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430746 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106380102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248444 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238819 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248444 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0217827.2_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline