Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00809672_s1
          See other NDUFA4 GE Assays ›
          Gene Symbol:
          Ndufa4
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4
          Gene Aliases:
          MLRQ
          Chromosome Location:
          Chr.6: 11900373 - 11907446 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00809672_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufa4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010886.2 NP_035016.1 3 261 112
          GenBank mRNA AK005084.1 - 3 266 112
          AK008357.1 - 3 258 112
          AK008506.1 - 3 252 112
          AK076036.1 - 3 262 112
          BC011114.1 - 3 224 3483028 112
          U59509.1 - 3 159 112

          Back To Top

          More Information


          Updated Mapping Information

          Based on assay re-evaluation in August 2010 with NCBI transcript and genome annotations (Genome Build NCBIM37.1), this assay is now annotated to be across exon-exon junction.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufa4

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          oxidation-reduction process
          hydrogen ion transmembrane transport
          cytochrome-c oxidase activity
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24239732 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24239733 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24239731 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24239734 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552832 NM_010886.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552832 AK005084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552832 AK008357.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552832 AK008506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552832 AK076036.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552832 NM_010886.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552832 AK005084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552832 AK008357.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552832 AK008506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P552832 AK076036.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 116052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30754 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173806 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552832 NM_010886.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552832 AK005084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552832 AK008357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552832 AK008506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552832 AK076036.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552832 NM_010886.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552832 AK005084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552832 AK008357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552832 AK008506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P552832 AK076036.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24239736 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173806 - -
          Whole Mouse Genome G4122A A_52_P552832 NM_010886.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552832 AK005084.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552832 AK008357.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552832 AK008506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552832 AK076036.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552832 NM_010886.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552832 AK005084.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552832 AK008357.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552832 AK008506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552832 AK076036.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101500 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173806 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05748850 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05748856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05748851 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05390395 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05390385 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24239733 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24239732 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24239734 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24239731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24239790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05145474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05011636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05748850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05030007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05320858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05320859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05390397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05600207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05605108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05748856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05390384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05390395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24239770 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10543135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543137 - -
          Affymetrix HT MG-430 PM Array Plate 1424085_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10543134 - -
          Mouse Genome 430 2.0 Array 1424085_PM_at - -
          Mouse Genome 430 2.0 Array 1424085_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17992_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17992_at - -
          Mouse Expression Set 430 1424085_at - -
          Mouse Expression Set 430 1424085_PM_at - -
          Murine Genome U74v2 Set 160477_at - -
          Murine Genome U74v2 Set 160477_PM_at - -
          Mouse Genome 430A 2.0 Array 1424085_PM_at - -
          Mouse Genome 430A 2.0 Array 1424085_at - -
          Affymetrix HT Mouse Genome 430A Array 1424085_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424085_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6860364 - -
          Illumina mouse-6 v1.0 expression beadchip 6860364 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860364 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2666062 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2666062 - -
          mouse-6 v1.1 (Illumina) scl017992.3_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 6860364 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl017992.3_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860364 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2666061 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2666061 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl017992.3_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline