Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00833860_g1
          See other EGFBP2 GE Assays ›
          Gene Symbol:
          Egfbp2
          Gene Name:
          epidermal growth factor binding protein type B
          Gene Aliases:
          Egfbp-2, Klk1b26, mGk-13
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00833860_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Egfbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010115.6 NP_034245.3 4 - 5 658 157
          GenBank mRNA X58628.1 - 4 - 5 658 157

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000787.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Egfbp2

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          zymogen activation
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005956 V00828.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005956 NM_010115.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005956 X58628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005956 V00828.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005956 NM_010115.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005956 X58628.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416647 NM_010115.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416647 X58628.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085324 - -
          Whole Mouse Genome G4122A A_51_P416647 NM_010115.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416647 X58628.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103081 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107727 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05925887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06064187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06064190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05913814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06064188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06398955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05913811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05913815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085319 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552579 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552580 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552575 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552577 - -
          Affymetrix HT MG-430 PM Array Plate 1421788_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552557 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552574 - -
          Mouse Genome 430 2.0 Array 1421788_PM_x_at - -
          Mouse Genome 430 2.0 Array 1421788_x_at - -
          Affymetrix Mouse Clariom S Array PSR0700006384.mm.1 TC0700000787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006384.mm.1 NM_010644.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006387.mm.1 TC0700000787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006387.mm.1 NM_010644.3 Refseq
          Mouse Expression Set 430 1421788_PM_x_at - -
          Mouse Expression Set 430 1421788_x_at - -
          Murine Genome U74v2 Set 100334_f_at - -
          Murine Genome U74v2 Set 161637_PM_f_at - -
          Murine Genome U74v2 Set 161637_f_at - -
          Murine Genome U74v2 Set 100334_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1421788_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1421788_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1421788_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1421788_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102190722 - -
          Illumina mouse-6 v1.0 expression beadchip 102190722 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246569 - -
          mouse-6 v1.1 (Illumina) GI_24475912-S - -
          Illumina Sentrix mouse-6.v1 bead array 102190722 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102190722 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2791241 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2791241 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline