Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00833976_s1
          See other IFNA5 GE Assays ›
          Gene Symbol:
          Ifna5
          Gene Name:
          interferon alpha 5
          Gene Aliases:
          Ifa5
          Chromosome Location:
          Chr.4: 88835525 - 88836094 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00833976_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifna5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010505.2 NP_034635.2 1 351 90
          GenBank mRNA BC120910.1 - 1 351 40091152 90
          BC120911.1 - 1 351 40091153 90

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifna5

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          cytokine activity
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030194 NM_010505.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030194 BC120910.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030194 BC120911.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030194 NM_010505.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030194 BC120910.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030194 BC120911.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P436401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092451 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127149 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136949 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03341968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03260395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092433 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10505880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514291 - -
          Affymetrix HT MG-430 PM Array Plate 1450614_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10505888 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505892 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505879 - -
          Mouse Genome 430 2.0 Array 1450614_PM_x_at - -
          Mouse Genome 430 2.0 Array 1450614_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15968_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15968_PM_at - -
          Mouse Expression Set 430 1422404_x_at - -
          Mouse Expression Set 430 1422403_at - -
          Mouse Expression Set 430 1450614_PM_x_at - -
          Mouse Expression Set 430 1450593_at - -
          Mouse Expression Set 430 1450614_x_at - -
          Mouse Expression Set 430 1450593_PM_at - -
          Mouse Expression Set 430 1422403_PM_at - -
          Mouse Expression Set 430 1422404_PM_x_at - -
          Murine Genome U74v2 Set 101619_at - -
          Murine Genome U74v2 Set 101791_PM_f_at - -
          Murine Genome U74v2 Set 96419_PM_f_at - -
          Murine Genome U74v2 Set 102149_PM_f_at - -
          Murine Genome U74v2 Set 101791_f_at - -
          Murine Genome U74v2 Set 102149_f_at - -
          Murine Genome U74v2 Set 102148_f_at - -
          Murine Genome U74v2 Set 102148_PM_f_at - -
          Murine Genome U74v2 Set 96419_f_at - -
          Murine Genome U74v2 Set 101619_PM_at - -
          Mouse Genome 430A 2.0 Array 1450593_PM_at - -
          Mouse Genome 430A 2.0 Array 1450593_at - -
          Mouse Genome 430A 2.0 Array 1422403_PM_at - -
          Mouse Genome 430A 2.0 Array 1422404_x_at - -
          Mouse Genome 430A 2.0 Array 1450614_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1450614_x_at - -
          Mouse Genome 430A 2.0 Array 1422403_at - -
          Mouse Genome 430A 2.0 Array 1422404_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1450614_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1450614_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 902058 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4780072 - -
          Illumina mouse-6 v1.0 expression beadchip 101240347 - -
          Illumina mouse-6 v1.0 expression beadchip 4780072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780072 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253698 - -
          mouse-6 v1.1 (Illumina) scl015968.1_325-S - -
          mouse-6 v1.1 (Illumina) GI_6754295-S - -
          Illumina Sentrix mouse-6.v1 bead array 101240347 - -
          Illumina Sentrix mouse-6.v1 bead array 4780072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015968.1_325-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101240347 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220596 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253698 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2648399 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220596 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline