Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00834740_sH
          See other MAGEA8 GE Assays ›
          Gene Symbol:
          Magea8
          Gene Name:
          melanoma antigen, family A, 8
          Gene Aliases:
          Mage-a8
          Chromosome Location:
          Chr.X: 154985775 - 154995850 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00834740_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Magea8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020020.4 NP_064404.1 4 559 217
          GenBank mRNA AK145480.1 - 4 560 217
          BC119510.2 - 1 250 40055160 217

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003230.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Magea8

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058786 NM_020020.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058786 AK145480.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058786 NM_020020.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058786 AK145480.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09465405 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09465411 - -
          Whole Mouse Genome G4122A A_52_P297142 NM_020020.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297142 AK145480.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297142 BC119510.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09465411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32758057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14996556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32758057 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10607441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607440 - -
          Affymetrix Mouse Gene 1.1 ST Array 10607454 - -
          Affymetrix Mouse Gene 1.1 ST Array 10607437 - -
          Affymetrix Mouse Gene 1.1 ST Array 10607443 - -
          Affymetrix Mouse Clariom S Array PSR0X00020211.mm.1 NM_020020.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020211.mm.1 TC0X00003230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00020210.mm.1 NM_020020.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020210.mm.1 TC0X00003230.mm.1 MTA TC
          Mouse Expression Set 430 1450542_PM_s_at - -
          Mouse Expression Set 430 1450542_s_at - -
          Mouse Genome 430A 2.0 Array 1450542_s_at - -
          Mouse Genome 430A 2.0 Array 1450542_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 891890 - -
          ABI Mouse Genome Survey Microarray 902627 - -
          ABI Mouse Genome Survey Microarray 903529 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1090577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2770293 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2770293 - -
          mouse-6 v1.1 (Illumina) scl0017144.1_460-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0017144.1_460-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090577 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2687002 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0017144.1_460-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline