Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00835439_g1
          See other UBE2C GE Assays ›
          Gene Symbol:
          Ube2c
          Gene Name:
          ubiquitin-conjugating enzyme E2C
          Gene Aliases:
          1110015A16Rik, D2Ertd695e
          Chromosome Location:
          Chr.2: 164769929 - 164772902 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00835439_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ube2c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026785.2 NP_081061.1 1 - 2 155 98
          GenBank mRNA AK003722.1 - 1 - 2 156 98
          AK136186.1 - 1 - 2 156 98
          AK160740.1 - 1 - 2 187 98
          BC147032.1 - 1 - 2 136 9056036 98
          BC147033.1 - 1 - 2 136 9056038 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ube2c

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          ubiquitin-dependent protein catabolic process
          cell cycle
          mitotic nuclear division
          negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
          exit from mitosis
          free ubiquitin chain polymerization
          protein ubiquitination
          regulation of mitotic metaphase/anaphase transition
          anaphase-promoting complex-dependent catabolic process
          positive regulation of exit from mitosis
          cell division
          protein K48-linked ubiquitination
          protein K11-linked ubiquitination
          nucleotide binding
          ubiquitin-protein transferase activity
          ATP binding
          ligase activity
          ubiquitin protein ligase binding
          ubiquitin protein ligase activity
          ubiquitin conjugating enzyme activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21819631 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21819633 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996941 AK136186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996941 AK160740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996941 BC147032.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996941 BC147033.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996941 BC171983.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451151 AK136186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451151 AK160740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451151 BC147032.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451151 BC147033.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451151 BC171983.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451151 AK136186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451151 AK160740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451151 BC147032.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451151 BC147033.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P451151 BC171983.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845389 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996941 AK136186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996941 AK160740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996941 BC147032.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996941 BC147033.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996941 BC171983.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451151 AK136186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451151 AK160740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451151 BC147032.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451151 BC147033.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451151 BC171983.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451151 AK136186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451151 AK160740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451151 BC147032.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451151 BC147033.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P451151 BC171983.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819632 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04772943 - -
          Whole Mouse Genome G4122A A_51_P451151 AK136186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451151 AK160740.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451151 BC147032.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451151 BC147033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451151 BC171983.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451151 AK136186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451151 AK160740.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451151 BC147032.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451151 BC147033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P451151 BC171983.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109429 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133021 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02219409 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02219407 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02219408 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01559111 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21819633 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21819631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01559111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01674561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02047824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02219397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02047812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02219398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02219410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819645 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10478573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478577 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478576 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478578 - -
          Affymetrix HT MG-430 PM Array Plate 1452954_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10478572 - -
          Mouse Genome 430 2.0 Array 1452954_PM_at - -
          Mouse Genome 430 2.0 Array 1452954_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68612_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68612_at - -
          Affymetrix Mouse Clariom S Array PSR0200020690.mm.1 NM_026785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020690.mm.1 NM_026785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020690.mm.1 TC0200002532.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020689.mm.1 NM_026785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020689.mm.1 NM_026785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020689.mm.1 TC0200002532.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020693.mm.1 NM_026785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020693.mm.1 NM_026785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020693.mm.1 TC0200002532.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020686.mm.1 NM_026785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020686.mm.1 NM_026785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020686.mm.1 TC0200002532.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020691.mm.1 NM_026785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020691.mm.1 NM_026785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020691.mm.1 TC0200002532.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020692.mm.1 NM_026785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020692.mm.1 NM_026785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020692.mm.1 TC0200002532.mm.1 MTA TC
          Mouse Expression Set 430 1452954_at - -
          Mouse Expression Set 430 1452954_PM_at - -
          Murine Genome U74v2 Set 161137_PM_r_at - -
          Murine Genome U74v2 Set 161137_r_at - -
          Mouse Genome 430A 2.0 Array 1452954_PM_at - -
          Mouse Genome 430A 2.0 Array 1452954_at - -
          Affymetrix HT Mouse Genome 430A Array 1452954_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452954_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 560131 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6130017 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130017 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229321 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229321 - -
          mouse-6 v1.1 (Illumina) scl068612.6_90-S - -
          Illumina Sentrix mouse-6.v1 bead array 6130017 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068612.6_90-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130017 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229321 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229321 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline