Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836721_s1
          See other OLFR1269 GE Assays ›
          Gene Symbol:
          Olfr1269
          Gene Name:
          olfactory receptor 1269
          Gene Aliases:
          MOR228-3
          Chromosome Location:
          Chr.2: 90118667 - 90119596 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836721_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1269
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146342.1 NP_666454.1 1 437 221
          GenBank mRNA BC116956.1 - 1 458 40126275 221
          BC116958.1 - 1 458 40126277 221

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1269

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P135864 BC116958.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P135864 NM_146343.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P135864 NM_146342.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P135864 BC116956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135864 BC116958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135864 NM_146343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135864 NM_146342.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135864 BC116956.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01800040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02110315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01228444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01828573 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484864 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484867 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484863 - -
          Affymetrix Mouse Clariom S Array PSR0200034125.mm.1 NM_146342.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034125.mm.1 NM_146342.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034125.mm.1 TC0200004080.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2320079 - -
          Illumina mouse-6 v1.0 expression beadchip 2320079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2738845 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2738845 - -
          mouse-6 v1.1 (Illumina) scl0258339.1_42-S - -
          Illumina Sentrix mouse-6.v1 bead array 2320079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258339.1_42-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320079 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2738845 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2738845 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline