Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836787_s1
          See other OLFR127 GE Assays ›
          Gene Symbol:
          Olfr127
          Gene Name:
          olfactory receptor 127
          Gene Aliases:
          MOR218-7
          Chromosome Location:
          Chr.17: 37903548 - 37904519 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836787_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr127
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146377.1 NP_666489.1 1 126 96
          GenBank mRNA BC132433.1 - 1 152 40130710 96
          BC145997.1 - 1 152 40131300 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr127

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P112080 NM_146377.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P112080 BC132433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P112080 BC145997.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P112080 NM_146377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P112080 BC132433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P112080 BC145997.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379318 NM_146377.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379318 BC132433.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379318 BC145997.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170335 - -
          Whole Mouse Genome G4122A A_51_P379318 NM_146377.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379318 BC132433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379318 BC145997.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112080 NM_146377.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112080 BC132433.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112080 BC145997.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13096182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13096184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13096186 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13229361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13493284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13553477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13579736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13163453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13579734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13096184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13431343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13482218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170368 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450907 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445170 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445168 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445165 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450906 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445169 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445167 - -
          Affymetrix Mouse Clariom S Array PSR1700007753.mm.1 NM_146377.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007753.mm.1 NM_146377.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007753.mm.1 TC1700000789.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007752.mm.1 TC1700000789.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 644586 - -
          ABI Mouse Genome Survey Microarray 811721 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2690180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690180 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2589097 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2589097 - -
          mouse-6 v1.1 (Illumina) scl0258374.1_284-S - -
          Illumina Sentrix mouse-6.v1 bead array 2690180 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258374.1_284-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690180 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2589097 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589097 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline