Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836830_s1
          See other OLFR462 GE Assays ›
          Gene Symbol:
          Olfr462
          Gene Name:
          olfactory receptor 462
          Gene Aliases:
          MOR240-3
          Chromosome Location:
          Chr.11: 87888959 - 87889894 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836830_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr462
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146411.2 NP_666523.2 1 883 180
          GenBank mRNA BC108939.1 - 883 40041186 180
          BC108940.1 - 884 180

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr462

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086797 BC108939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086797 BC108940.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086797 NM_146411.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086797 BC108939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086797 BC108940.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086797 NM_146411.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P441855 BC106970.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P441855 BC108939.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P441855 BC108940.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P441855 NM_146413.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P441855 BC106969.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996849 - -
          Whole Mouse Genome G4122A A_52_P441855 BC106970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P441855 BC108939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P441855 BC108940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P441855 NM_146413.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P441855 BC106969.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100018 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136358 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09443105 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09094501 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09443098 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09443112 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09443103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09443098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09554751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09610406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09554753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09443104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09443103 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10389671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389669 - -
          Affymetrix Mouse Gene 1.1 ST Array 10389668 - -
          Affymetrix Mouse Gene 1.1 ST Array 10389670 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1170441 - -
          Illumina mouse-6 v1.0 expression beadchip 1170441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2770285 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2770285 - -
          mouse-6 v1.1 (Illumina) scl0258406.1_75-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258406.1_75-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2875857 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2770285 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2875857 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline