Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837382_s1
          See other OLFR1501 GE Assays ›
          Gene Symbol:
          Olfr1501
          Gene Name:
          olfactory receptor 1501
          Gene Aliases:
          MOR212-3
          Chromosome Location:
          Chr.19: 13838224 - 13839171 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837382_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1501
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146633.2 NP_666844.1 1 550 168

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1501

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262124 NM_146633.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262124 NM_146633.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262124 NM_146633.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262124 NM_146633.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P262124 NM_146633.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P262124 NM_146633.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31966073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31966074 - -
          Whole Mouse Genome G4122A A_52_P262124 NM_146633.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P262124 NM_146633.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109416 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14331849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966080 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466371 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466370 - -
          Affymetrix Mouse Clariom S Array PSR1900010886.mm.1 NM_146633.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010886.mm.1 NM_146633.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010886.mm.1 TC1900001190.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 796608 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6770193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2768174 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2768174 - -
          mouse-6 v1.1 (Illumina) scl0258626.1_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770193 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258626.1_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770193 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2803864 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803864 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2768174 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline