Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837426_s1
          See other OLFR1154 GE Assays ›
          Gene Symbol:
          Olfr1154
          Gene Name:
          olfactory receptor 1154
          Gene Aliases:
          MOR173-2
          Chromosome Location:
          Chr.2: 87902742 - 87903674 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837426_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1154
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146647.2 NP_666858.2 1 449 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1154

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039746 NM_146647.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58920 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039746 NM_146647.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P104258 NM_146647.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P104258 NM_146647.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01911648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01834105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01911650 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484698 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484697 - -
          Affymetrix Mouse Clariom S Array PSR0200033897.mm.1 NM_146647.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033897.mm.1 NM_146647.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033897.mm.1 TC0200003997.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894163 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102760176 - -
          Illumina mouse-6 v1.0 expression beadchip 4480082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004480082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228004 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228004 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241483 - -
          mouse-6 v1.1 (Illumina) GI_33239201-S - -
          mouse-6 v1.1 (Illumina) scl0258641.1_158-S - -
          Illumina Sentrix mouse-6.v1 bead array 4480082 - -
          Illumina Sentrix mouse-6.v1 bead array 102760176 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258641.1_158-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4480082 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102760176 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228004 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228004 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline