Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837464_s1
          See other OLFR441 GE Assays ›
          Gene Symbol:
          Olfr441
          Gene Name:
          olfactory receptor 441
          Gene Aliases:
          MOR261-3
          Chromosome Location:
          Chr.6: 43115744 - 43116676 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837464_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr441
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146655.1 NP_666866.1 1 370 195

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr441

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506250 NM_146655.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P506250 NM_146655.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506250 NM_146655.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P506250 NM_146655.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P506250 NM_146655.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P506250 NM_146655.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24401613 - -
          Whole Mouse Genome G4122A A_52_P506250 NM_146655.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P506250 NM_146655.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100361 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05553465 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05225548 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05553464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05118528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05812369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05022283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05812367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05225548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05320458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05330901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05320457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05664754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05022275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05320450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401625 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537818 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537817 - -
          Affymetrix Mouse Clariom S Array PSR0600003763.mm.1 TC0600000479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003763.mm.1 NM_146655.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003763.mm.1 NM_146655.1 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3360450 - -
          MouseWG-6 v2 Expression BeadChip 60091 - -
          Illumina mouse-6 v1.0 expression beadchip 3360450 - -
          Illumina mouse-6 v1.0 expression beadchip 60091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000060091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2751563 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1260140 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2751563 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260140 - -
          mouse-6 v1.1 (Illumina) scl0258649.1_47-S - -
          mouse-6 v1.1 (Illumina) scl00258649.1_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 3360450 - -
          Illumina Sentrix mouse-6.v1 bead array 60091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258649.1_47-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258649.1_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360450 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 60091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1260140 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2751563 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1260140 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2751563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline