Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837547_s1
          See other OLFR816 GE Assays ›
          Gene Symbol:
          Olfr816
          Gene Name:
          olfactory receptor 816
          Gene Aliases:
          MOR113-1
          Chromosome Location:
          Chr.10: 129911338 - 129912276 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837547_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr816
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146672.1 NP_666883.1 1 147 183
          GenBank mRNA BC100682.1 - 1 145 183

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr816

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114704 NM_146672.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114704 BC100682.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114704 NM_146672.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114704 BC100682.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P522849 NM_146672.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P522849 BC100682.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547741 - -
          Whole Mouse Genome G4122A A_52_P522849 NM_146672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P522849 BC100682.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122954 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08560388 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08876450 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08876447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08560393 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08560385 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08560387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08882340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08608325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08608323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547736 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373656 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373655 - -
          Affymetrix Mouse Clariom S Array PSR1000023972.mm.1 NM_146672.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023972.mm.1 NM_146672.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023972.mm.1 TC1000003196.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 423256 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6620026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006620026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2731239 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2731239 - -
          mouse-6 v1.1 (Illumina) scl0258667.1_137-S - -
          Illumina Sentrix mouse-6.v1 bead array 6620026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258667.1_137-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6620026 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2731239 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2731239 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline