Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837751_s1
          See other OLFR583 GE Assays ›
          Gene Symbol:
          Olfr583
          Gene Name:
          olfactory receptor 583
          Gene Aliases:
          MOR14-6
          Chromosome Location:
          Chr.7: 103051300 - 103052259 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837751_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr583
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146757.1 NP_666968.1 1 739 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr583

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P248673 NM_146757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P248673 NM_146757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P248673 NM_146757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P248673 NM_146757.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P248673 NM_146757.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P248673 NM_146757.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381794 - -
          Whole Mouse Genome G4122A A_52_P248673 NM_146757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P248673 NM_146757.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104960 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05946737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06329692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06591717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381855 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555761 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555760 - -
          Affymetrix Mouse Clariom S Array PSR0700012713.mm.1 TC0700001475.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012713.mm.1 NM_146757.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012713.mm.1 NM_146757.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 318279 - -
          ABI Mouse Genome Survey Microarray 872370 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4230692 - -
          Illumina mouse-6 v1.0 expression beadchip 4230692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737050 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737050 - -
          mouse-6 v1.1 (Illumina) scl0258752.1_118-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258752.1_118-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2737050 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2737050 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0