Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837995_s1
          See other OLFR1219 GE Assays ›
          Gene Symbol:
          Olfr1219
          Gene Name:
          olfactory receptor 1219
          Gene Aliases:
          MOR233-6
          Chromosome Location:
          Chr.2: 89074154 - 89075089 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837995_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1219
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146899.2 NP_667110.2 1 677 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1219

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2100982 NM_146899.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100982 NM_146899.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P163901 NM_146899.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163901 NM_146899.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01947105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01947117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01447736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01683259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01553016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01947119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01953652 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484784 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484783 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 895375 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4780168 - -
          Illumina mouse-6 v1.0 expression beadchip 4780168 - -
          Illumina mouse-6 v1.0 expression beadchip 106130280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780168 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2773151 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256546 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2773151 - -
          mouse-6 v1.1 (Illumina) GI_22129084-S - -
          mouse-6 v1.1 (Illumina) scl0258901.1_329-S - -
          Illumina Sentrix mouse-6.v1 bead array 4780168 - -
          Illumina Sentrix mouse-6.v1 bead array 106130280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258901.1_329-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106130280 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780168 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2940661 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2940661 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline