Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840153_m1
          See other ART2B GE Assays ›
          Gene Symbol:
          Art2b
          Gene Name:
          ADP-ribosyltransferase 2b
          Gene Aliases:
          ART2.2, ARTC2, Art, Ly92b, Rt-6, Rt6, Rt6-2
          Chromosome Location:
          Chr.7: 101575729 - 101585583 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840153_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Art2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019915.2 NP_064299.2 2 - 3 787 76
          GenBank mRNA AF016463.1 - 2 - 3 787 76
          AF016465.1 - 2 - 3 787 76
          X87612.1 - 2 - 3 787 76

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Art2b

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ADP-ribosylation
          peptidyl-arginine ADP-ribosylation
          NAD catabolic process
          NAD(P)+-protein-arginine ADP-ribosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          hydrolase activity, acting on glycosyl bonds

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964886 AF016463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964886 AF016465.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964886 X87612.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964886 NM_019915.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240650 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128945 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964886 AF016463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964886 AF016465.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964886 X87612.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964886 NM_019915.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505372 AF016463.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505372 AF016465.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505372 NM_019915.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25374331 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128945 - -
          Whole Mouse Genome G4122A A_51_P505372 AF016463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505372 AF016465.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505372 NM_019915.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137241 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128945 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06293245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06455825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05994723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06107982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05994726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06293254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06552437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06293245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10565995 - -
          Affymetrix HT MG-430 PM Array Plate 1420794_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10565994 - -
          Mouse Genome 430 2.0 Array 1420794_PM_at - -
          Mouse Genome 430 2.0 Array 1420794_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11872_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11872_PM_at - -
          Mouse Expression Set 430 1420794_PM_at - -
          Mouse Expression Set 430 1420794_at - -
          Murine Genome U74v2 Set 99402_at - -
          Murine Genome U74v2 Set 99402_PM_at - -
          Mouse Genome 430A 2.0 Array 1420794_PM_at - -
          Mouse Genome 430A 2.0 Array 1420794_at - -
          Affymetrix HT Mouse Genome 430A Array 1420794_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420794_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 848291 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1850064 - -
          Illumina mouse-6 v1.0 expression beadchip 1850064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850064 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215074 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215074 - -
          mouse-6 v1.1 (Illumina) scl30962.3.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 1850064 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30962.3.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215074 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215074 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline