Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840197_m1
          See other CST10 GE Assays ›
          Gene Symbol:
          Cst10
          Gene Name:
          cystatin 10 (chondrocytes)
          Gene Aliases:
          DD72
          Chromosome Location:
          Chr.2: 149405249 - 149410278 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840197_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cst10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021405.2 NP_067380.1 2 - 3 474 104
          GenBank mRNA AB036743.1 - 2 - 3 474 104
          BC048364.1 - 2 - 3 390 4920219 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002210.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cst10

          Gene Ontology Categories:

          Function(s) Process(es)

          chondrocyte differentiation
          negative regulation of endopeptidase activity
          cell maturation
          cysteine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959858 BC048364.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959858 AB036743.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959858 NM_021405.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544209 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150813 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959858 BC048364.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959858 AB036743.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959858 NM_021405.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21734563 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00524727 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150813 - -
          Whole Mouse Genome G4122A A_51_P227999 BC048364.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227999 AB036743.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227999 NM_021405.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139231 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150813 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02262675 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01712280 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01712278 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02262677 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02262676 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02262682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21734536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01919474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01633688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01712284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02177078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02177077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02262677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01919472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01845426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02030638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21734538 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10476932 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476934 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476933 - -
          Affymetrix HT MG-430 PM Array Plate 1449447_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10476931 - -
          Mouse Genome 430 2.0 Array 1449447_PM_at - -
          Mouse Genome 430 2.0 Array 1449447_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58214_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58214_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200017452.mm.1 NM_021405.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017452.mm.1 NM_021405.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017452.mm.1 TC0200002210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017449.mm.1 NM_021405.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017449.mm.1 NM_021405.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017449.mm.1 TC0200002210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017453.mm.1 NM_021405.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017453.mm.1 NM_021405.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017453.mm.1 TC0200002210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017451.mm.1 NM_021405.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017451.mm.1 NM_021405.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017451.mm.1 TC0200002210.mm.1 MTA TC
          Mouse Expression Set 430 1449447_at - -
          Mouse Expression Set 430 1449447_PM_at - -
          Mouse Genome 430A 2.0 Array 1449447_at - -
          Mouse Genome 430A 2.0 Array 1449447_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449447_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449447_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 570297 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 630279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000630279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2711743 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2711743 - -
          mouse-6 v1.1 (Illumina) scl20150.4.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 630279 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20150.4.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 630279 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2711743 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2711743 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline