Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840228_m1
          See other 9530002B09RIK GE Assays ›
          Gene Symbol:
          9530002B09Rik
          Gene Name:
          RIKEN cDNA 9530002B09 gene
          Gene Aliases:
          Aump, Vpp1, Vppl
          Chromosome Location:
          Chr.4: 122689259 - 122705135 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840228_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9530002B09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023865.3 NP_076354.1 4 - 5 520 112
          GenBank mRNA AF319955.1 - 4 - 5 505 112
          AK020534.1 - 4 - 5 470 112
          AY077734.1 - 4 - 5 499 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001381.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 9530002B09Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          protein depalmitoylation
          receptor-mediated endocytosis
          nervous system development
          palmitoyl-(protein) hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971129 NM_023865.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735846 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87743 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44266 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118057 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162224 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971129 NM_023865.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23259002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23258995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23259004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23258991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23258989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23258986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23258993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23259006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23258988 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05382660 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162224 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118057 - -
          Whole Mouse Genome G4122A A_51_P502521 AY077734.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502521 NM_023865.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502521 AF319955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502521 AK020534.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127615 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118057 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162224 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03341014 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03341016 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03599644 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03341007 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03599641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23258989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23258986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23258992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23258988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23259008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03703729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23259002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23259017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03755594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23259021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23259008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23258998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03755597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23258986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23259011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03703726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03341007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23259006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23258988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23259009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23258993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23259015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03814997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03599644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23259007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23259014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10507778 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507782 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507779 - -
          Affymetrix HT MG-430 PM Array Plate 1419242_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10507777 - -
          Mouse Genome 430 2.0 Array 1419242_PM_at - -
          Mouse Genome 430 2.0 Array 1419242_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77432_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77432_at - -
          Affymetrix Mouse Clariom S Array PSR0400010395.mm.1 NM_023865.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010395.mm.1 NM_023865.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010395.mm.1 TC0400001381.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400010392.mm.1 NM_023865.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010392.mm.1 NM_023865.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010392.mm.1 TC0400001381.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400010397.mm.1 NM_023865.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010397.mm.1 NM_023865.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010397.mm.1 TC0400001381.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400010391.mm.1 NM_023865.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010391.mm.1 NM_023865.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010391.mm.1 TC0400001381.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400010393.mm.1 NM_023865.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010393.mm.1 NM_023865.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010393.mm.1 TC0400001381.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400010394.mm.1 NM_023865.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010394.mm.1 NM_023865.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010394.mm.1 TC0400001381.mm.1 MTA TC
          Mouse Expression Set 430 1419242_PM_at - -
          Mouse Expression Set 430 1419242_at - -
          Mouse Genome 430A 2.0 Array 1419242_at - -
          Mouse Genome 430A 2.0 Array 1419242_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419242_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419242_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 538570 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1500576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230372 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230372 - -
          mouse-6 v1.1 (Illumina) scl25006.5.1_146-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25006.5.1_146-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230372 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230372 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline