Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840330_g1
          See other LIPT2 GE Assays ›
          Gene Symbol:
          Lipt2
          Gene Name:
          lipoyl(octanoyl) transferase 2 (putative)
          Gene Aliases:
          2610209A20Rik
          Chromosome Location:
          Chr.7: 100159277 - 100160931 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840330_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lipt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026010.2 NP_080286.2 1 - 2 475 130
          GenBank mRNA AK008917.1 - 1 - 2 456 130
          AK011925.1 - 1 - 2 475 130
          BC115472.1 - 1 - 2 475 40100448 130
          BC115473.1 - 1 - 2 470 40100449 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001400.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lipt2

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein modification process
          biological_process
          lipoate biosynthetic process
          protein lipoylation
          protein binding
          octanoyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          ligase activity
          lipoyl(octanoyl) transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366032 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366033 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366029 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366026 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130777 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044463 AK011925.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25365999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25366012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25365997 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06299058 - -
          Whole Mouse Genome G4122A A_52_P421196 AK013127.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P421196 BE648757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P421196 NM_026010.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P421196 AK008917.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P421196 AK011925.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119711 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06299058 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05984512 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06428102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06428101 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06428097 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366029 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366027 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366033 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366032 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25366026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25365981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25366012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06158127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06434678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05930179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06428101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06245420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05990701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05955136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05927803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05927805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06428097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05927797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05955135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25365995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06354159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06211695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25366009 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555294 - -
          Affymetrix HT MG-430 PM Array Plate 1423357_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10555293 - -
          Mouse Genome 430 2.0 Array 1423357_PM_at - -
          Mouse Genome 430 2.0 Array 1423357_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67164_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67164_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700011869.mm.1 TC0700001400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011869.mm.1 NM_026010.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011869.mm.1 NM_026010.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011871.mm.1 TC0700001400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011871.mm.1 NM_026010.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011871.mm.1 NM_026010.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011870.mm.1 TC0700001400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011870.mm.1 NM_026010.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011870.mm.1 NM_026010.2 Refseq
          Mouse Expression Set 430 1423357_at - -
          Mouse Expression Set 430 1423357_PM_at - -
          Mouse Genome 430A 2.0 Array 1423357_PM_at - -
          Mouse Genome 430A 2.0 Array 1423357_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 652915 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4670600 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670600 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744228 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744228 - -
          mouse-6 v1.1 (Illumina) scl32321.2_292-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670600 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32321.2_292-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670600 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2744228 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3145474 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2744228 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3145474 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline