Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840378_m1
          See other CELA3B GE Assays ›
          Gene Symbol:
          Cela3b
          Gene Name:
          chymotrypsin-like elastase family, member 3B
          Gene Aliases:
          0910001F22Rik, 2310074F01Rik, AI504000, Ela3, Ela3b
          Chromosome Location:
          Chr.4: 137421008 - 137430520 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840378_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cela3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026419.2 NP_080695.1 6 - 7 664 77
          GenBank mRNA AK008858.1 - 6 - 7 663 77
          AK009129.1 - 6 - 7 663 77
          AK010149.1 - 6 - 7 664 77
          BC056210.1 - 5 - 6 603 77
          BC061066.1 - 6 - 7 664 30309250 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003808.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cela3b

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042535 AK010149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042535 BC056210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042535 BC061066.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042535 AK009129.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042535 NM_026419.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488052 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123813 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042535 AK010149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042535 BC056210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042535 BC061066.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042535 AK009129.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042535 NM_026419.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P340456 AK010149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P340456 BC056210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P340456 BC061066.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P340456 AK009129.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P340456 NM_026419.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23339766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23339771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23339744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23339761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23339758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23339760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23339775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23339759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23339770 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123813 - -
          Whole Mouse Genome G4122A A_51_P340456 AK010149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340456 BC056210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340456 BC061066.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340456 AK009129.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P340456 NM_026419.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108876 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123813 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03255709 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03255708 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03757630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03336596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23339719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03615871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03355564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03757630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03255709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03633604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03604842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03355565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03325412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03255708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23339727 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10517577 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517575 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517576 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517574 - -
          Affymetrix HT MG-430 PM Array Plate 1435611_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1415884_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1415883_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10517573 - -
          Mouse Genome 430 2.0 Array 1442170_PM_at - -
          Mouse Genome 430 2.0 Array 1442170_at - -
          Mouse Genome 430 2.0 Array 1435611_PM_x_at - -
          Mouse Genome 430 2.0 Array 1415884_PM_at - -
          Mouse Genome 430 2.0 Array 1415883_PM_a_at - -
          Mouse Genome 430 2.0 Array 1435611_x_at - -
          Mouse Genome 430 2.0 Array 1435012_x_at - -
          Mouse Genome 430 2.0 Array 1415884_at - -
          Mouse Genome 430 2.0 Array 1415883_a_at - -
          Mouse Genome 430 2.0 Array 1435012_PM_x_at - -
          Mouse Genome 430 2.0 Array 1437326_PM_x_at - -
          Mouse Genome 430 2.0 Array 1437326_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67868_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67868_at - -
          Affymetrix Mouse Clariom S Array PSR0400031410.mm.1 TC0400003808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031410.mm.1 NM_026419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031410.mm.1 NM_026419.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031409.mm.1 TC0400003808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031409.mm.1 NM_026419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031409.mm.1 NM_026419.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031405.mm.1 TC0400003808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031405.mm.1 NM_026419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031405.mm.1 NM_026419.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031411.mm.1 TC0400003808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031411.mm.1 NM_026419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031411.mm.1 NM_026419.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031413.mm.1 TC0400003808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031413.mm.1 NM_026419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031413.mm.1 NM_026419.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031408.mm.1 TC0400003808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031408.mm.1 NM_026419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031408.mm.1 NM_026419.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031412.mm.1 TC0400003808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031412.mm.1 NM_026419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031412.mm.1 NM_026419.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031406.mm.1 TC0400003808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031406.mm.1 NM_026419.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031406.mm.1 NM_026419.2 Refseq
          Mouse Expression Set 430 1415883_a_at - -
          Mouse Expression Set 430 1415883_PM_a_at - -
          Mouse Expression Set 430 1435611_PM_x_at - -
          Mouse Expression Set 430 1437326_PM_x_at - -
          Mouse Expression Set 430 1415884_PM_at - -
          Mouse Expression Set 430 1415884_at - -
          Mouse Expression Set 430 1435012_PM_x_at - -
          Mouse Expression Set 430 1435012_x_at - -
          Mouse Expression Set 430 1437326_x_at - -
          Mouse Expression Set 430 1435611_x_at - -
          Mouse Genome 430A 2.0 Array 1435611_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1437326_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1435012_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1415883_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1437326_x_at - -
          Mouse Genome 430A 2.0 Array 1415884_at - -
          Mouse Genome 430A 2.0 Array 1435611_x_at - -
          Mouse Genome 430A 2.0 Array 1435012_x_at - -
          Mouse Genome 430A 2.0 Array 1415883_a_at - -
          Mouse Genome 430A 2.0 Array 1415884_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415884_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435611_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1415884_at - -
          Affymetrix HT Mouse Genome 430A Array 1435611_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1415883_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415883_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 710369 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3990711 - -
          Illumina mouse-6 v1.0 expression beadchip 4060731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003990711 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258024 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2689363 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258024 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2689363 - -
          mouse-6 v1.1 (Illumina) scl0067868.1_42-S - -
          mouse-6 v1.1 (Illumina) scl067868.2_48-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060731 - -
          Illumina Sentrix mouse-6.v1 bead array 3990711 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067868.1_42-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067868.2_48-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060731 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3990711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2671137 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2671137 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067868.2_48-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline