Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840980_m1
          See other BC025446 GE Assays ›
          Gene Symbol:
          BC025446
          Gene Name:
          cDNA sequence BC025446
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 75216338 - 75222661 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840980_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC025446
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC025446.1 - 3 - 4 453 5053382 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000554.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339154 BC025446.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339154 BC025446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339154 BC025446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339154 BC025446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339154 BC025446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339154 BC025446.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12060768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11946539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12424858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12060764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12192017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12060749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12147036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12288404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11946540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11852061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11852062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30341298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10424690 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424687 - -
          Affymetrix HT MG-430 PM Array Plate 1425712_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10424686 - -
          Mouse Genome 430 2.0 Array 1425712_at - -
          Mouse Genome 430 2.0 Array 1425712_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223631_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223631_at - -
          Affymetrix Mouse Clariom S Array PSR1500003789.mm.1 TC1500000554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500003792.mm.1 TC1500000554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500003791.mm.1 TC1500000554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500003790.mm.1 TC1500000554.mm.1 MTA TC
          Mouse Expression Set 430 1425712_PM_at - -
          Mouse Expression Set 430 1425712_at - -
          Mouse Genome 430A 2.0 Array 1425712_PM_at - -
          Mouse Genome 430A 2.0 Array 1425712_at - -
          Affymetrix HT Mouse Genome 430A Array 1425712_at - -
          Affymetrix HT Mouse Genome 430A Array 1425712_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 842635 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6590050 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240955 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240955 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2915835 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2915833 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2915835 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2915833 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline