Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00842185_g1
          See other RASD1 GE Assays ›
          Gene Symbol:
          Rasd1
          Gene Name:
          RAS, dexamethasone-induced 1
          Gene Aliases:
          Dexras1
          Chromosome Location:
          Chr.11: 59963181 - 59965080 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00842185_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rasd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009026.4 NP_033052.1 1 - 2 429 144
          GenBank mRNA AF009246.1 - 1 - 2 433 144
          AK038932.1 - 1 - 2 432 144
          BC034166.1 - 1 - 2 399 4989312 144
          CT010376.1 - 1 - 2 292 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002865.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rasd1

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          nitric oxide mediated signal transduction
          small GTPase mediated signal transduction
          negative regulation of transcription, DNA-templated
          nucleotide binding
          protein binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839977 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839989 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839979 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839981 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839985 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839987 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839984 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839988 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839978 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496726 AF009246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496726 AK038932.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496726 BC034166.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496726 NM_009026.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496726 AF009246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496726 AK038932.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496726 BC034166.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496726 NM_009026.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613815 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109061 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496726 AF009246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496726 AK038932.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496726 BC034166.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496726 NM_009026.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496726 AF009246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496726 AK038932.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496726 BC034166.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496726 NM_009026.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P496726 AF009246.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P496726 AK038932.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P496726 BC034166.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P496726 NM_009026.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P496726 AF009246.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P496726 AK038932.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P496726 BC034166.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P496726 NM_009026.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27840019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27839990 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109061 - -
          Whole Mouse Genome G4122A A_52_P496726 AF009246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496726 AK038932.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496726 BC034166.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496726 NM_009026.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496726 AF009246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496726 AK038932.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496726 BC034166.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496726 NM_009026.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109297 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124449 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109061 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09330582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09330595 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09330589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09330587 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09330584 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09330566 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09330567 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839989 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839977 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839979 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839984 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839985 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839988 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839978 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839987 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27839981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27839998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27840012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09330590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09330586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09499653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09330566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09618339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09618331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09330594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09330587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27839990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27840000 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10386457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386458 - -
          Affymetrix HT MG-430 PM Array Plate 1423619_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10386455 - -
          Mouse Genome 430 2.0 Array 1423619_at - -
          Mouse Genome 430 2.0 Array 1423619_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19416_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19416_at - -
          Affymetrix Mouse Clariom S Array PSR1100026011.mm.1 NM_009026.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026011.mm.1 TC1100002865.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100026013.mm.1 NM_009026.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026013.mm.1 TC1100002865.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100026012.mm.1 NM_009026.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026012.mm.1 TC1100002865.mm.1 MTA TC
          Mouse Expression Set 430 1423619_at - -
          Mouse Expression Set 430 1423619_PM_at - -
          Murine Genome U74v2 Set 99032_at - -
          Murine Genome U74v2 Set 99032_PM_at - -
          Mouse Genome 430A 2.0 Array 1423619_at - -
          Mouse Genome 430A 2.0 Array 1423619_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423619_at - -
          Affymetrix HT Mouse Genome 430A Array 1423619_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 662485 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2450463 - -
          Illumina mouse-6 v1.0 expression beadchip 2450463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002450463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2647195 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2647195 - -
          mouse-6 v1.1 (Illumina) scl40151.2_453-S - -
          Illumina Sentrix mouse-6.v1 bead array 2450463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40151.2_453-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2647197 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2647197 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40151.2_453-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline