Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00847504_s1
          See other OLFR107 GE Assays ›
          Gene Symbol:
          Olfr107
          Gene Name:
          olfactory receptor 107
          Gene Aliases:
          MOR156-3
          Chromosome Location:
          Chr.17: 37405477 - 37406509 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00847504_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr107
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146511.2 NP_666722.2 1 521 177
          GenBank mRNA BC141327.1 - 1 521 9055939 177
          BC141329.1 - 1 521 9055941 177
          BC148321.1 - 1 448 40054936 177

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr107

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605596 NM_146511.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605596 BC141327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605596 BC141329.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605596 BC148321.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605596 NM_146511.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605596 BC141327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605596 BC141329.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605596 BC148321.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24013 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162802 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605596 NM_146511.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605596 BC141327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605596 BC141329.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605596 BC148321.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605596 NM_146511.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605596 BC141327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605596 BC141329.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605596 BC148321.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P605596 NM_146511.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P605596 BC141327.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P605596 BC141329.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P605596 BC148321.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P605596 NM_146511.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P605596 BC141327.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P605596 BC141329.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P605596 BC148321.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168562 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162802 - -
          Whole Mouse Genome G4122A A_52_P605596 NM_146511.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605596 BC141327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605596 BC141329.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605596 BC148321.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605596 NM_146511.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605596 BC141327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605596 BC141329.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605596 BC148321.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100501 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162802 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149923 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149924 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13575886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13575891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13462346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13242361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13462184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13242364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13388957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13149918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13242366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13192784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13517539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13462343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13213405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168618 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445134 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445133 - -
          Affymetrix Mouse Clariom S Array PSR1700007718.mm.1 NM_146511.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007718.mm.1 NM_146511.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007718.mm.1 TC1700000772.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 836695 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 360338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247423 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247423 - -
          mouse-6 v1.1 (Illumina) scl50730.1.1_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 360338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50730.1.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247423 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247423 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline