Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00847580_s1
          See other OLFR935 GE Assays ›
          Gene Symbol:
          Olfr935
          Gene Name:
          olfactory receptor 935
          Gene Aliases:
          MOR171-11
          Chromosome Location:
          Chr.9: 38994507 - 38995433 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00847580_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr935
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146746.1 NP_666957.1 1 640 144

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr935

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P173976 NM_146746.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P173976 NM_146746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P173976 NM_146746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P173976 NM_146746.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P173976 NM_146746.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P173976 NM_146746.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436934 - -
          Whole Mouse Genome G4122A A_51_P173976 NM_146746.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173976 NM_146746.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104690 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07640396 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07640426 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07640408 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07640422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07749616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07749609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436948 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592385 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592384 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 732622 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4010021 - -
          Illumina mouse-6 v1.0 expression beadchip 4010021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2667577 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2667577 - -
          mouse-6 v1.1 (Illumina) scl6624.1.1_36-S - -
          Illumina Sentrix mouse-6.v1 bead array 4010021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6624.1.1_36-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2667577 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2667577 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline