Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00848660_s1
          See other OLFR1216 GE Assays ›
          Gene Symbol:
          Olfr1216
          Gene Name:
          olfactory receptor 1216
          Gene Aliases:
          MOR233-9
          Chromosome Location:
          Chr.2: 89013127 - 89014062 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00848660_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1216
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146893.2 NP_667104.2 1 48 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004036.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1216

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365844 NM_146893.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365844 NM_146893.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102550 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365844 NM_146893.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365844 NM_146893.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P365844 NM_146893.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P365844 NM_146893.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102550 - -
          Whole Mouse Genome G4122A A_51_P365844 NM_146893.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365844 NM_146893.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103085 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102608 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21412109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01861330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02142382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21412167 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484778 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484777 - -
          Affymetrix Mouse Clariom S Array PSR0200033996.mm.1 NM_146893.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033996.mm.1 NM_146893.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033996.mm.1 TC0200004036.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894428 - -
          ABI Mouse Genome Survey Microarray 902143 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4230026 - -
          Illumina mouse-6 v1.0 expression beadchip 4230026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2743091 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2743091 - -
          mouse-6 v1.1 (Illumina) scl19028.1.1_37-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19028.1.1_37-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230026 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2743091 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2743091 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0