Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00850017_g1
          See other H2AFB1 GE Assays ›
          Gene Symbol:
          H2afb1
          Gene Name:
          H2A histone family, member B1
          Gene Aliases:
          1700113O17Rik, H2A.Bbd, H2al2, TSEG-1, Tseg1
          Chromosome Location:
          Chr.2: 17996422 - 17996946 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00850017_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2afb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026627.1 NP_080903.1 1 265 104
          GenBank mRNA AK006586.1 - 1 251 104
          AK015538.1 - 1 256 104
          AK018839.1 - 1 265 104
          AK018883.1 - 1 187 104
          AK018922.1 - 1 251 104
          AK018951.1 - 1 251 104
          AK018962.1 - 1 251 104
          AK160415.1 - 1 251 104
          BC048736.1 - 1 229 104
          BC061062.1 - 1 277 6774311 104
          EU079024.1 - 1 192 104

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2afb1

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P119810 AK018839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P119810 AK018883.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P119810 AK018922.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P119810 AK018951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P119810 AK018962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P119810 AK018839.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P119810 AK018883.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P119810 AK018922.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P119810 AK018951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P119810 AK018962.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303147 AK018839.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303147 AK018883.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303147 AK018922.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303147 AK018951.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303147 AK018962.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21037661 - -
          Whole Mouse Genome G4122A A_51_P303147 AK018839.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303147 AK018883.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303147 AK018922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303147 AK018951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303147 AK018962.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01573533 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01202616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21037695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01853910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01986106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02111587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01961799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02153679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01919257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01719063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01961800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01202616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02153685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01560129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21037648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01627410 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608197 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608034 - -
          Affymetrix HT MG-430 PM Array Plate 1418303_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608196 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608033 - -
          Affymetrix Mouse Gene 1.1 ST Array 10480317 - -
          Mouse Genome 430 2.0 Array 1418303_at - -
          Mouse Genome 430 2.0 Array 1418303_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68231_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68231_PM_at - -
          Mouse Expression Set 430 1418303_PM_at - -
          Mouse Expression Set 430 1418303_at - -
          Mouse Genome 430A 2.0 Array 1418303_PM_at - -
          Mouse Genome 430A 2.0 Array 1418303_at - -
          Affymetrix HT Mouse Genome 430A Array 1418303_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418303_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 927861 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1580300 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2689749 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2689749 - -
          mouse-6 v1.1 (Illumina) scl19641.1.15_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 1580300 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19641.1.15_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2689749 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2689749 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline