Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01136596_m1
          See other RNF121 GE Assays ›
          Gene Symbol:
          Rnf121
          Gene Name:
          ring finger protein 121
          Gene Aliases:
          4930544L10Rik
          Chromosome Location:
          Chr.7: 102019145 - 102072428 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01136596_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf121
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029211.2 NP_083487.2 6 - 7 629 72
          GenBank mRNA AK034806.1 - 6 - 7 1943 72
          AK078509.1 - 6 - 7 590 72
          AK159032.1 - 6 - 7 766 72
          AK168821.1 - 6 - 7 642 72
          AK169089.1 - 6 - 7 642 72
          BC022686.1 - 6 - 7 629 4241984 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003851.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf121

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein ubiquitination
          ER-associated ubiquitin-dependent protein catabolic process
          endoplasmic reticulum unfolded protein response
          molecular_function
          zinc ion binding
          metal ion binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700032528.mm.1 NM_029211.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032528.mm.1 NM_029211.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032528.mm.1 TC0700003851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032533.mm.1 NM_029211.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032533.mm.1 NM_029211.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032533.mm.1 TC0700003851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032539.mm.1 NM_029211.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032539.mm.1 NM_029211.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032539.mm.1 TC0700003851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032527.mm.1 NM_029211.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032527.mm.1 NM_029211.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032527.mm.1 TC0700003851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032525.mm.1 NM_029211.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032525.mm.1 NM_029211.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032525.mm.1 TC0700003851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032530.mm.1 NM_029211.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032530.mm.1 NM_029211.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032530.mm.1 TC0700003851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032531.mm.1 NM_029211.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032531.mm.1 NM_029211.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032531.mm.1 TC0700003851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032534.mm.1 NM_029211.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032534.mm.1 NM_029211.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032534.mm.1 TC0700003851.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 525060 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline