Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01137059_m1
          See other CADPS2 GE Assays ›
          Gene Symbol:
          Cadps2
          Gene Name:
          Ca2+-dependent activator protein for secretion 2
          Gene Aliases:
          A230044C21Rik, Caps-2, Caps2, Cpd2
          Chromosome Location:
          Chr.6: 23262773 - 23839421 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01137059_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cadps2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252105.1 NP_001239034.1 1 - 2 633 74
          NM_001252106.1 NP_001239035.1 1 - 2 633 74
          NM_001252107.1 NP_001239036.1 1 - 2 633 74
          NM_001252108.1 NP_001239037.1 1 - 2 633 74
          NM_001252109.1 NP_001239038.1 1 - 2 633 74
          NM_001252110.1 NP_001239039.1 1 - 2 633 74
          NM_153163.4 NP_694803.3 1 - 2 633 74
          GenBank mRNA AB098623.1 - 1 - 2 632 74
          AB291946.1 - 1 - 2 633 74
          AB291947.1 - 1 - 2 448 74
          AB291948.1 - 1 - 2 518 74
          AB291949.1 - 1 - 2 487 74
          AB291950.1 - 1 - 2 518 74
          AF000969.2 - 1 - 2 261 74
          AK042634.1 - 1 - 2 475 74
          AK046596.1 - 1 - 2 479 74
          AK163203.1 - 1 - 2 513 74
          AY072800.1 - 2 - 3 262 74
          BC047394.1 - 1 - 2 261 74

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF000969.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cadps2

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          exocytosis
          cellular response to starvation
          protein transport
          synaptic vesicle priming
          positive regulation of exocytosis
          protein binding
          lipid binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 TC0600001931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252105.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252105.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252107.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252107.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252108.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252108.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252109.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252109.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252110.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_001252110.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_153163.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015166.mm.1 NM_153163.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 TC0600001931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252105.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252105.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252107.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252107.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252108.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252108.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252109.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252109.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252110.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_001252110.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_153163.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015159.mm.1 NM_153163.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 TC0600001931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252105.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252105.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252107.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252107.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252108.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252108.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252109.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252109.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252110.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_001252110.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_153163.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015164.mm.1 NM_153163.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 TC0600001931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252105.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252105.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252107.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252107.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252108.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252108.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252109.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252109.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252110.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_001252110.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_153163.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015174.mm.1 NM_153163.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 TC0600001931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252105.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252105.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252107.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252107.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252108.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252108.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252109.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252109.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252110.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_001252110.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_153163.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015161.mm.1 NM_153163.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 TC0600001931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252105.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252105.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252107.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252107.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252108.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252108.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252109.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252109.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252110.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_001252110.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_153163.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015173.mm.1 NM_153163.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 TC0600001931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252105.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252105.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252107.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252107.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252108.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252108.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252109.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252109.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252110.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_001252110.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_153163.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015162.mm.1 NM_153163.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 TC0600001931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252105.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252105.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252107.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252107.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252108.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252108.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252109.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252109.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252110.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_001252110.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_153163.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015169.mm.1 NM_153163.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 TC0600001931.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252105.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252105.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252106.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252107.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252107.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252108.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252108.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252109.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252109.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252110.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_001252110.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_153163.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015171.mm.1 NM_153163.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687610 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline