Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01160771_s1
          See other OLFR263 GE Assays ›
          Gene Symbol:
          Olfr263
          Gene Name:
          olfactory receptor 263
          Gene Aliases:
          GA_x5J8B7TTCWC-858-415, IA3, MOR256-31, MOR256-37P, MOR256-61, Olfr263-ps1, Olfr42
          Chromosome Location:
          Chr.13: 21132777 - 21133730 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01160771_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr263
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010984.1 NP_035114.1 1 326 98
          GenBank mRNA AF102516.1 - 1 137 98
          BC116918.1 - 1 405 40126237 98
          BC120698.1 - 1 405 40129621 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000160.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr263

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094787 BC116918.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094787 BC120698.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094787 NM_010984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094787 AF102516.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094787 BC116918.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094787 BC120698.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094787 NM_010984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094787 AF102516.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281950 BC116918.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281950 BC120698.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281950 NM_010984.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281950 AF102516.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P244520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871238 - -
          Whole Mouse Genome G4122A A_51_P244520 - -
          Whole Mouse Genome G4122A A_51_P281950 BC116918.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281950 BC120698.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281950 NM_010984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281950 AF102516.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10876832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10699423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871224 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403902 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403900 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403899 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403901 - -
          Affymetrix Mouse Clariom S Array PSR1300001080.mm.1 NM_010984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001080.mm.1 NM_010984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001080.mm.1 TC1300000160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001079.mm.1 TC1300000160.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 767591 - -
          ABI Mouse Genome Survey Microarray 897904 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2060037 - -
          Illumina mouse-6 v1.0 expression beadchip 2060037 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002060037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2592893 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2592893 - -
          mouse-6 v1.1 (Illumina) scl018341.1_205-S - -
          Illumina Sentrix mouse-6.v1 bead array 2060037 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018341.1_205-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060037 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2592893 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2592893 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline