Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01165081_m1
          See other NUP205 GE Assays ›
          Gene Symbol:
          Nup205
          Gene Name:
          nucleoporin 205
          Gene Aliases:
          3830404O05Rik, AV248391, mKIAA0225
          Chromosome Location:
          Chr.6: 35177408 - 35247599 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01165081_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nup205
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027513.1 NP_081789.1 13 - 14 2030 65
          GenBank mRNA AK014419.1 - 13 - 14 2030 65
          AK129093.1 - 13 - 14 2215 65
          BC058729.1 - 12 - 13 1999 65
          BC060139.1 - 10 - 11 1543 65
          BC060711.1 - 10 - 11 1543 65
          BC141333.1 - 13 - 14 2216 9055945 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000344.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear pore complex assembly
          structural constituent of nuclear pore

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113698 BC060531.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113698 BC060711.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113698 BC027832.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113698 BC037401.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113698 BC058729.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252492 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 374349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570888 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441212 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424870 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 711037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 712643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 574969 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753118 AK028763.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753118 NM_001081165.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 429556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500311 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 146018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 158792 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159093 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139340 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147868 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159623 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113698 BC060531.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113698 BC060711.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113698 BC027832.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113698 BC037401.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113698 BC058729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177062 AK014419.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P275233 BC060531.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P275233 BC060711.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P275233 BC141333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P275233 BC027832.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P275233 BC037401.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05693656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05443467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05818224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05114587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05707049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05072779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05072777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05624092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05361691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05081542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05693661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05823898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05264639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05071054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05818219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05072775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05866463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24360156 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537281 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537252 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537286 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537266 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537262 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537265 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537285 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537258 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537248 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537276 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537267 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537271 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537259 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537250 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537249 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537261 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537280 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537256 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537260 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537253 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537272 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537277 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537270 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537257 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537269 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537283 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537255 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537268 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537254 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537264 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537278 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537282 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537273 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537263 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537251 - -
          Affymetrix HT MG-430 PM Array Plate 1454743_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430343_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10537246 - -
          Mouse Genome 430 2.0 Array 1454743_PM_at - -
          Mouse Genome 430 2.0 Array 1454743_at - -
          Mouse Genome 430 2.0 Array 1430343_at - -
          Mouse Genome 430 2.0 Array 1430343_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70699_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70699_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600002605.mm.1 TC0600000344.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600002605.mm.1 NM_027513.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002605.mm.1 NM_027513.1 Refseq
          Mouse Expression Set 430 1454743_PM_at - -
          Mouse Expression Set 430 1430343_PM_at - -
          Mouse Expression Set 430 1430343_at - -
          Mouse Expression Set 430 1454743_at - -
          Affymetrix HT Mouse Genome 430B Array 1430343_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430343_at - -
          Affymetrix HT Mouse Genome 430B Array 1454743_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454743_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 691280 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2602855 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2602855 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238382 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline