Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01165579_m1
          See other MRS2 GE Assays ›
          Gene Symbol:
          Mrs2
          Gene Name:
          MRS2 magnesium homeostasis factor homolog (S. cerevisiae)
          Gene Aliases:
          Gm902, HPT, Mrs2l, RPT
          Chromosome Location:
          Chr.13: 24987483 - 25020358 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01165579_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrs2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013389.2 NP_001013407.2 9 - 10 1153 71
          GenBank mRNA AK136100.1 - 4 - 5 444 71
          BB015377.2 - 2 - 3 254 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001672.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrs2

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          biological_process
          magnesium ion transport
          molecular_function
          magnesium ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28890273 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28890271 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618187 NM_001013389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618187 AK136100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618187 NM_001013389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618187 AK136100.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 422661 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640892 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166688 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618187 NM_001013389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618187 AK136100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618187 NM_001013389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618187 AK136100.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28890278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07813660 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07813701 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02648694 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07813690 - -
          Whole Mouse Genome G4122A A_52_P618187 NM_001013389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P618187 AK136100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P618187 NM_001013389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P618187 AK136100.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07813690 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10636349 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10636352 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10598056 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10636351 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28890273 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28890271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28890180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10558519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10727456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10636354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10638176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10619262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10619265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10835537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10603039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10495156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10477528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10486547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11111930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10598055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10603030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10619278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10486553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10486546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28890297 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10408355 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408353 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408354 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408351 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408356 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408349 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408350 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408352 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408358 - -
          Affymetrix HT MG-430 PM Array Plate 1435548_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10408348 - -
          Mouse Genome 430 2.0 Array 1435548_at - -
          Mouse Genome 430 2.0 Array 1435548_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 380836_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 380836_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300010414.mm.1 NM_001013389.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010414.mm.1 TC1300001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010413.mm.1 NM_001013389.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010413.mm.1 TC1300001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010415.mm.1 NM_001013389.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010415.mm.1 TC1300001672.mm.1 MTA TC
          Mouse Expression Set 430 1435548_at - -
          Mouse Expression Set 430 1435548_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435548_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435548_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 883789 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253039 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2868102 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2868102 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline