Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01166782_s1
          See other D17H6S53E GE Assays ›
          Gene Symbol:
          D17H6S53E
          Gene Name:
          DNA segment, Chr 17, human D6S53E
          Gene Aliases:
          G4, NG34
          Chromosome Location:
          Chr.17: 35126402 - 35128855 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01166782_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D17H6S53E
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033477.2 NP_258438.1 3 843 76
          GenBank mRNA BC138707.1 - 1 539 8861729 76
          BC138708.1 - 1 539 8861730 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene D17H6S53E

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157755 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157757 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275751 BC138708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275751 NM_033477.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275751 BC138707.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275751 BC138708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275751 NM_033477.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275751 BC138707.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275751 BC138708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275751 NM_033477.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275751 BC138707.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275751 BC138708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275751 NM_033477.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275751 BC138707.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275751 BC138708.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275751 NM_033477.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275751 BC138707.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275751 BC138708.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275751 NM_033477.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P275751 BC138707.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157768 - -
          Whole Mouse Genome G4122A A_51_P275751 BC138708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275751 NM_033477.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275751 BC138707.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275751 BC138708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275751 NM_033477.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275751 BC138707.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13174129 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13174126 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13174127 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13174110 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13174119 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157757 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31157755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13192772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13174109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13174127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13174110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157768 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444718 - -
          Affymetrix Mouse Gene 1.1 ST Array 10444717 - -
          Affymetrix Mouse Clariom S Array PSR1700006784.mm.1 NM_033477.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006784.mm.1 NM_033477.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006784.mm.1 TC1700000669.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006785.mm.1 NM_033477.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006785.mm.1 NM_033477.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006785.mm.1 TC1700000669.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006788.mm.1 NM_033477.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006788.mm.1 NM_033477.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006788.mm.1 TC1700000669.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006789.mm.1 NM_033477.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006789.mm.1 NM_033477.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006789.mm.1 TC1700000669.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006787.mm.1 NM_033477.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006787.mm.1 NM_033477.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006787.mm.1 TC1700000669.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 607124 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161414 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161414 - -
          mouse-6 v1.1 (Illumina) GI_51591904-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161414 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161414 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline