Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01172197_m1
          See other 2310009A05RIK GE Assays ›
          Gene Symbol:
          2310009A05Rik
          Gene Name:
          RIKEN cDNA 2310009A05 gene
          Gene Aliases:
          5031420N21Rik
          Chromosome Location:
          Chr.9: 73039718 - 73042775 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01172197_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310009A05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK003016.1 - 1 - 2 141 87
          AK009244.1 - 1 - 2 148 87
          BC037369.1 - 1 - 2 115 87
          BC100521.1 - 1 - 2 140 87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY414798.1 - 1 - 2 61 Chr. 9 - 73039809 - 73042774 [+] on Build GRCm38 87
          GenBankmRNA AA269388.1 - 1 - 2 Chr. 9 - 73039740 - 73042761 [+] on Build GRCm38 86
          GenBankmRNA AA154652.1 - 1 - 2 Chr. 9 - 73039740 - 73042668 [+] on Build GRCm38 87
          GenBankmRNA W66785.1 - 134 Chr. - - [-] on Build 87
          GenBankmRNA AV511221.1 - 1 - 2 115 Chr. 9 - 73039758 - 73042753 [+] on Build GRCm38 87
          GenBankmRNA BY241316.1 - 1 - 2 150 Chr. 9 - 73039724 - 73042740 [+] on Build GRCm38 87
          GenBankmRNA AV004941.1 - 1 - 2 157 Chr. 9 - 73039741 - 73042773 [+] on Build GRCm38 87
          GenBankmRNA BY247632.1 - 1 - 2 150 Chr. 9 - 73039724 - 73042773 [+] on Build GRCm38 87
          GenBankmRNA AI181193.1 - 1 - 2 Chr. 9 - 73039757 - 73042760 [+] on Build GRCm38 85
          GenBankmRNA CJ321971.1 - 1 - 2 83 Chr. 9 - 73039787 - 73042773 [+] on Build GRCm38 87
          GenBankmRNA W82419.1 - Chr. - - [-] on Build 87
          GenBankmRNA AI615968.1 - 1 - 2 130 Chr. 9 - 73039740 - 73042772 [+] on Build GRCm38 87
          GenBankmRNA CJ185120.1 - 1 - 2 143 Chr. 9 - 73039730 - 73042688 [+] on Build GRCm38 87
          GenBankmRNA BY094141.1 - 1 - 2 141 Chr. 9 - 73039730 - 73042687 [+] on Build GRCm38 87
          GenBankmRNA EH106697.1 - 1 - 2 163 Chr. 9 - 73039718 - 73042779 [+] on Build GRCm38 87
          GenBankmRNA CJ315031.1 - 1 - 2 86 Chr. 9 - 73039784 - 73042773 [+] on Build GRCm38 87
          GenBankmRNA CF383640.1 - 1 - 2 168 Chr. 9 - 73039724 - 73042779 [+] on Build GRCm38 87
          GenBankmRNA BY083018.1 - 1 - 2 141 Chr. 9 - 73039730 - 73042666 [+] on Build GRCm38 87
          GenBankmRNA CN838569.1 - 1 - 2 143 Chr. 9 - 73039728 - 73042779 [+] on Build GRCm38 87
          GenBankmRNA AI037776.1 - 1 - 2 Chr. 9 - 73039793 - 73042774 [+] on Build GRCm38 85
          GenBankmRNA AA444725.1 - 1 - 2 113 Chr. 9 - 73039756 - 73042719 [+] on Build GRCm38 86
          GenBankmRNA BX524967.1 - 1 - 2 116 Chr. 9 - 73039755 - 73042779 [+] on Build GRCm38 87
          GenBankmRNA CJ178332.1 - 1 - 2 143 Chr. 9 - 73039730 - 73042772 [+] on Build GRCm38 87
          GenBankmRNA BY240366.1 - 1 - 2 150 Chr. 9 - 73039724 - 73042718 [+] on Build GRCm38 87
          GenBankmRNA AI152819.1 - 1 - 2 Chr. 9 - 73039753 - 73042774 [+] on Build GRCm38 85
          GenBankmRNA DV040541.1 - 193 Chr. - - [-] on Build 87
          GenBankmRNA AA272667.1 - 1 - 2 166 Chr. 9 - 73039718 - 73042754 [+] on Build GRCm38 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phosphatidylinositol glycan anchor biosynthesis,
          class B
          Pigb Pig-b -31.0 Anti-sense Chr. 9 - 73013726 - 73039699 on NCBI Build GRCm38
          Downstream RAB27A,
          member RAS oncogene family
          Rab27a 2210402C08Rik,
          2410003M20Rik,
          4933437C11Rik,
          ash
          2.0kb Sense Chr. 9 - 73044810 - 73097623 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 210130 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109526 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129218 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129218 - -
          Whole Mouse Genome G4122A A_52_P203852 AK003016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P203852 AK009244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P203852 AV139032.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P203852 BC037369.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P203852 AI037776.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129218 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1435238_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1435237_PM_at - -
          Mouse Genome 430 2.0 Array 1435237_PM_at - -
          Mouse Genome 430 2.0 Array 1435237_at - -
          Mouse Genome 430 2.0 Array 1435238_x_at - -
          Mouse Genome 430 2.0 Array 1435238_PM_x_at - -
          Mouse Expression Set 430 1435237_at - -
          Mouse Expression Set 430 1435238_PM_x_at - -
          Mouse Expression Set 430 1435237_PM_at - -
          Mouse Expression Set 430 1435238_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1435238_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1435238_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1435237_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435237_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 556632 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0