Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175872_m1
          See other AB124611 GE Assays ›
          Gene Symbol:
          AB124611
          Gene Name:
          cDNA sequence AB124611
          Gene Aliases:
          Gm1706
          Chromosome Location:
          Chr.9: 21526166 - 21545331 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175872_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AB124611
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001198794.1 NP_001185723.1 6 - 7 653 94
          NM_206536.2 NP_996259.2 5 - 6 563 94
          GenBank mRNA AB124611.1 - 6 - 7 553 94
          AB124612.1 - 5 - 6 463 94
          AK137593.1 - 7 - 8 740 94
          AK154442.1 - 6 - 7 640 94
          AK162293.1 - 6 - 7 640 94
          AK168311.1 - 6 - 7 640 94
          AK170200.1 - 6 - 7 640 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000199.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AB124611

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066778 AK168311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066778 AK170200.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066778 BU696592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066778 NM_206536.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066778 AB124611.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129691 AK168311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129691 AK170200.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129691 NM_206536.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129691 AK137593.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129691 AK162293.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630311 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 110875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375223 AK008492.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375223 AK081629.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375223 AK088941.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375223 BC028640.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375223 BC067028.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375223 NM_028099.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58080 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141338 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166043 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139532 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066778 AK168311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066778 AK170200.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066778 BU696592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066778 NM_206536.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066778 AB124611.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129691 AK168311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129691 AK170200.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129691 NM_206536.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129691 AK137593.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129691 AK162293.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347588 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141338 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166043 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01882291 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105271 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141338 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08096798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08096801 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07464573 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08096796 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08096793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07707261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07973472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07585091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07509077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07707262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07574068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07593152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08096795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07939804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08096791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07593154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07801885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347614 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583675 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583674 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583672 - -
          Affymetrix HT MG-430 PM Array Plate 1438475_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10583669 - -
          Mouse Genome 430 2.0 Array 1438475_PM_at - -
          Mouse Genome 430 2.0 Array 1438475_at - -
          Affymetrix Mouse Clariom S Array PSR0900001434.mm.1 NM_001198794.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001434.mm.1 NM_001198794.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001434.mm.1 NM_206536.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001434.mm.1 NM_206536.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001434.mm.1 TC0900000199.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001433.mm.1 NM_001198794.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001433.mm.1 NM_001198794.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001433.mm.1 NM_206536.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001433.mm.1 NM_206536.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001433.mm.1 TC0900000199.mm.1 MTA TC
          Mouse Expression Set 430 1438475_at - -
          Mouse Expression Set 430 1438475_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438475_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438475_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 474924 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2190180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002190180 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2719139 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2719139 - -
          mouse-6 v1.1 (Illumina) scl0382062.2_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 2190180 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0382062.2_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190180 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2719139 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2719139 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline