Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01178036_s1
          See other OLFR1351 GE Assays ›
          Gene Symbol:
          Olfr1351
          Gene Name:
          olfactory receptor 1351
          Gene Aliases:
          MOR139-4
          Chromosome Location:
          Chr.10: 79017324 - 79018283 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01178036_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1351
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147040.1 NP_667251.1 1 902 136
          GenBank mRNA BC104112.1 - 1 902 40043939 136
          BC104113.1 - 1 901 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1351

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358303 NM_147040.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358303 BC104112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358303 BC104113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358303 NM_147040.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358303 BC104112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358303 BC104113.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104000 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358303 NM_147040.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358303 BC104112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358303 BC104113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358303 NM_147040.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358303 BC104112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358303 BC104113.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358303 NM_147040.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358303 BC104112.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358303 BC104113.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358303 NM_147040.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358303 BC104112.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358303 BC104113.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27276222 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104000 - -
          Whole Mouse Genome G4122A A_51_P358303 NM_147040.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358303 BC104112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358303 BC104113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358303 NM_147040.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358303 BC104112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358303 BC104113.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116577 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104000 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08675337 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08675336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27276196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08675336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08851470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08345686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27276174 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364427 - -
          Affymetrix Mouse Gene 1.1 ST Array 10364426 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 853045 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3060075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060075 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247058 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247058 - -
          mouse-6 v1.1 (Illumina) scl38718.1.1_293-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38718.1.1_293-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247058 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247058 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline