Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01181781_m1
          See other REG2 GE Assays ›
          Gene Symbol:
          Reg2
          Gene Name:
          regenerating islet-derived 2
          Gene Aliases:
          RegII
          Chromosome Location:
          Chr.6: 78405155 - 78408097 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01181781_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Reg2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009043.1 NP_033069.1 4 - 5 416 88
          GenBank mRNA AK008196.1 - 4 - 5 415 88
          BC098503.1 - 3 - 4 399 6431377 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000877.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Reg2

          Gene Ontology Categories:

          Function(s) Process(es)

          suckling behavior
          myelination
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333549 NM_009043.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333549 AK008196.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333549 BC098503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333549 NM_009043.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333549 AK008196.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333549 BC098503.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 258225 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68161 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165979 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333549 NM_009043.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333549 AK008196.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333549 BC098503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333549 NM_009043.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333549 AK008196.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333549 BC098503.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569972 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165979 - -
          Whole Mouse Genome G4122A A_51_P333549 NM_009043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333549 AK008196.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333549 BC098503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333549 NM_009043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333549 AK008196.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333549 BC098503.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138091 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165979 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05588706 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05418876 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05588703 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05418875 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05588725 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05418878 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05588705 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05588710 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05588702 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05588716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24570010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05397549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05677630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05588714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05677637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05588717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05588702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05588705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05677636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05051416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24570002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569983 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10539195 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539196 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539197 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539198 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539199 - -
          Affymetrix HT MG-430 PM Array Plate 1416139_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10539194 - -
          Mouse Genome 430 2.0 Array 1416139_at - -
          Mouse Genome 430 2.0 Array 1416139_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19693_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19693_at - -
          Affymetrix Mouse Clariom S Array PSR0600006680.mm.1 TC0600000877.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600006680.mm.1 NM_009043.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006680.mm.1 NM_009043.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006679.mm.1 TC0600000877.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600006679.mm.1 NM_009043.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006679.mm.1 NM_009043.1 Refseq
          Mouse Expression Set 430 1416139_PM_at - -
          Mouse Expression Set 430 1416139_at - -
          Murine Genome U74v2 Set 161302_PM_r_at - -
          Murine Genome U74v2 Set 161302_r_at - -
          Murine Genome U74v2 Set 95786_at - -
          Murine Genome U74v2 Set 95786_PM_at - -
          Murine Genome U74v2 Set 165408_PM_f_at - -
          Murine Genome U74v2 Set 165408_f_at - -
          Mouse Genome 430A 2.0 Array 1416139_at - -
          Mouse Genome 430A 2.0 Array 1416139_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416139_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416139_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 668430 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2470072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2612268 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2612268 - -
          mouse-6 v1.1 (Illumina) scl019693.5_25-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl019693.5_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2612268 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2612268 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline