Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01182148_m1
          See other TEX37 GE Assays ›
          Gene Symbol:
          Tex37
          Gene Name:
          testis expressed 37
          Gene Aliases:
          1700011F03Rik
          Chromosome Location:
          Chr.6: 70913089 - 70951150 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01182148_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex37
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028825.3 NP_083101.1 3 - 4 213 79
          GenBank mRNA AK005862.1 - 3 - 4 214 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002456.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tex37

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P190630 NM_028825.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P190630 AK005862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P190630 BC049736.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P190630 NM_028825.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P190630 AK005862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P190630 BC049736.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178896 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100668 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P190630 NM_028825.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P190630 AK005862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P190630 BC049736.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P190630 NM_028825.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P190630 AK005862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P190630 BC049736.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24533723 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100668 - -
          Whole Mouse Genome G4122A A_52_P190630 NM_028825.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P190630 AK005862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P190630 BC049736.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P190630 NM_028825.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P190630 AK005862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P190630 BC049736.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100668 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05487372 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04998930 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05094554 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05487368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24533709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05123465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05507133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05411441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05282181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05123459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05123460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05123473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05527343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05527344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05282183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05507130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05123458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05487367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05282184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05527342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05487366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05527345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05002900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05742577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05123468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05507128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24533671 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545270 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545268 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545267 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545269 - -
          Affymetrix HT MG-430 PM Array Plate 1430892_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545266 - -
          Mouse Genome 430 2.0 Array 1430892_at - -
          Mouse Genome 430 2.0 Array 1430892_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74221_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74221_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600018996.mm.1 TC0600002456.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018996.mm.1 NM_028825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018996.mm.1 NM_028825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018997.mm.1 TC0600002456.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018997.mm.1 NM_028825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018997.mm.1 NM_028825.3 Refseq
          Mouse Expression Set 430 1430892_at - -
          Mouse Expression Set 430 1430892_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430892_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430892_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 532019 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4120008 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2639308 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2639308 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2639308 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline