Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01182686_m1
          See other 1700054K19RIK GE Assays ›
          Gene Symbol:
          1700054K19Rik
          Gene Name:
          RIKEN cDNA 1700054K19 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 112210708 - 112212519 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01182686_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700054K19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027865.1 2 - 3 128 98

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130067 BY707289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130067 NR_027865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130067 AK006783.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130067 AK018861.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130067 AK018961.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697254 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774068 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153082 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130067 BY707289.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130067 NR_027865.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130067 AK006783.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130067 AK018861.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130067 AK018961.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05363390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05423811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05217453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05217448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05589204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05082916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05217450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05516424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05363386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05174327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05084032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05355878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05084037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05404075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05082920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05859689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05082918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05319494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05319493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05352084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05859690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05084034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05363385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05404076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05217451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24752518 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1440775_PM_at - -
          Mouse Genome 430 2.0 Array 1440775_at - -
          Mouse Genome 430 2.0 Array 1440775_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67336_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67336_PM_at - -
          Mouse Expression Set 430 1440775_PM_at - -
          Mouse Expression Set 430 1440775_at - -
          Murine Genome U74v2 Set 167505_at - -
          Murine Genome U74v2 Set 167505_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440775_at - -
          Affymetrix HT Mouse Genome 430B Array 1440775_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 930768 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102260180 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline