Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01182835_m1
          See other RNF181 GE Assays ›
          Gene Symbol:
          Rnf181
          Gene Name:
          ring finger protein 181
          Gene Aliases:
          1700047F16Rik, 2500002L14Rik, 6230415N01Rik, C77350
          Chromosome Location:
          Chr.6: 72359714 - 72366967 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01182835_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf181
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025607.3 NP_079883.3 1 - 2 136 88
          GenBank mRNA AK010854.1 - 1 - 2 149 88
          AK159788.1 - 1 - 2 149 88
          AK166904.1 - 1 - 2 148 88
          AK166995.1 - 1 - 2 149 88
          AK167396.1 - 1 - 2 149 88
          AK167633.1 - 1 - 2 149 88
          AK167935.1 - 1 - 2 149 88
          AK168670.1 - 1 - 2 150 88
          BC005559.1 - 1 - 2 160 88
          BC083119.1 - 1 - 2 137 5374213 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002486.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf181

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein autoubiquitination
          ubiquitin-protein transferase activity
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541329 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143915 BC005559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143915 BC083119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143915 AK168670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143915 AK207916.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143915 AK208815.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143915 BC005559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143915 BC083119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143915 AK168670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143915 AK207916.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143915 AK208815.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989384 BC005559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989384 BC083119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989384 AK168670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989384 AK195168.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989384 AK202921.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177985 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177899 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147956 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989384 BC005559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989384 BC083119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989384 AK168670.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989384 AK195168.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989384 AK202921.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143915 BC005559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143915 BC083119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143915 AK168670.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143915 AK207916.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143915 AK208815.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143915 BC005559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143915 BC083119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143915 AK168670.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143915 AK207916.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143915 AK208815.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541332 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147956 - -
          Whole Mouse Genome G4122A A_51_P143915 BC005559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143915 BC083119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143915 AK168670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143915 AK207916.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143915 AK208815.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143915 BC005559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143915 BC083119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143915 AK168670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143915 AK207916.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143915 AK208815.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139140 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05091229 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05033909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05219630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05091226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05776002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05868628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05425192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05033909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05091231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05075275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05091225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05033911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541343 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545395 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545396 - -
          Affymetrix HT MG-430 PM Array Plate 1453800_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423729_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545394 - -
          Mouse Genome 430 2.0 Array 1453800_at - -
          Mouse Genome 430 2.0 Array 1423729_PM_a_at - -
          Mouse Genome 430 2.0 Array 1453800_PM_at - -
          Mouse Genome 430 2.0 Array 1423729_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66510_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66510_at - -
          Affymetrix Mouse Clariom S Array PSR0600019256.mm.1 TC0600002486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019256.mm.1 NM_025607.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019258.mm.1 TC0600002486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019258.mm.1 NM_025607.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019259.mm.1 TC0600002486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019259.mm.1 NM_025607.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019260.mm.1 TC0600002486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019260.mm.1 NM_025607.4 Refseq
          Mouse Expression Set 430 1423729_a_at - -
          Mouse Expression Set 430 1423729_PM_a_at - -
          Mouse Expression Set 430 1453800_at - -
          Mouse Expression Set 430 1453800_PM_at - -
          Murine Genome U74v2 Set 101971_PM_at - -
          Murine Genome U74v2 Set 101971_at - -
          Mouse Genome 430A 2.0 Array 1423729_a_at - -
          Mouse Genome 430A 2.0 Array 1423729_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423729_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423729_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1453800_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453800_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 545160 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4670079 - -
          MouseWG-6 v2 Expression BeadChip 2120026 - -
          Illumina mouse-6 v1.0 expression beadchip 2120026 - -
          Illumina mouse-6 v1.0 expression beadchip 4670079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2606017 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2682063 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2606017 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2682063 - -
          mouse-6 v1.1 (Illumina) scl0001255.1_2-S - -
          mouse-6 v1.1 (Illumina) scl066510.1_267-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670079 - -
          Illumina Sentrix mouse-6.v1 bead array 2120026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066510.1_267-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001255.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670079 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120026 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2682063 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2606015 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2606015 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2682063 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066510.1_267-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0