Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01183193_m1
          See other SLC25A18 GE Assays ›
          Gene Symbol:
          Slc25a18
          Gene Name:
          solute carrier family 25 (mitochondrial carrier), member 18
          Gene Aliases:
          1500015I14Rik, AW125787
          Chromosome Location:
          Chr.6: 120773510 - 120794169 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01183193_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc25a18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081048.2 NP_001074517.1 6 - 7 488 69
          GenBank mRNA AK005250.1 - 4 - 5 400 69
          BC116828.1 - 6 - 7 488 40126147 69
          BC137603.1 - 6 - 7 488 8860622 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001354.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transporter secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          transmembrane transport
          molecular_function
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103225 AK005250.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103225 BC116828.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103225 NM_001081048.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103225 BC137603.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694906 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 419463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79769 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121901 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100140 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103225 AK005250.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103225 BC116828.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103225 NM_001081048.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103225 BC137603.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100140 - -
          Whole Mouse Genome G4122A A_51_P507664 AK005250.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P507664 BC116828.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P507664 NM_001081048.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P507664 BC137603.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24800322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05442451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05352905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05490079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05866859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05364849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05442215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05823130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05442447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05377449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05377441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05352904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05237975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05866876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05377442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05377448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05522805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05194971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05377450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05442446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05851265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05418586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05866865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05866863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24800194 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10541273 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541272 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541271 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541276 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541270 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541269 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541277 - -
          Affymetrix HT MG-430 PM Array Plate 1428964_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10541268 - -
          Mouse Genome 430 2.0 Array 1428964_PM_at - -
          Mouse Genome 430 2.0 Array 1428964_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71803_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71803_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600010805.mm.1 TC0600001354.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600010805.mm.1 NM_001081048.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010805.mm.1 NM_001081048.2 Refseq
          Mouse Expression Set 430 1428964_at - -
          Mouse Expression Set 430 1428964_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428964_at - -
          Affymetrix HT Mouse Genome 430B Array 1428964_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 633759 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2704027 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2704027 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2704027 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline