Search
Search

Clec1b| Gene Symbol: Clec1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204239.1 | NP_001191168.1 | 4 - 5 | 768 | 123 | |
| NM_001204253.1 | NP_001191182.1 | 3 - 4 | 616 | 123 | ||
| NM_019985.3 | NP_064369.1 | 5 - 6 | 864 | 123 | ||
| GenBank mRNA | AF201457.1 | - | 5 - 6 | 575 | 123 | |
| AK007925.1 | - | 5 - 6 | 844 | 123 | ||
| AK154961.1 | - | 119 | 123 | |||
| BC064054.1 | - | 5 - 6 | 864 | 574821 | 123 | |
| EF070191.1 | - | 4 - 5 | 458 | 123 | ||
| EF070192.1 | - | 3 - 4 | 303 | 123 | ||
Set Membership: |
|